Skip to Content
Merck
All Photos(1)

Key Documents

1.04095

Sigma-Aldrich

Glycerol

for fluorescence (microscopy)

Synonym(s):

1,2,3-Propanetriol, Glycerin, Glycerol, Protol, Trihydroxylpropane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NA.21
Pricing and availability is not currently available.

grade

for fluorescence (microscopy)

Quality Level

form

liquid

autoignition temp.

400 °C

expl. lim.

2.6-11.3 % V)

IVD

for in vitro diagnostic use

dilution

(histology)

pH range

5.5-8.0 ( in water)

bp

290 °C/1013 hPa (Decomposition)

transition temp

flash point 199 °C

density

1.261 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing

General description

Glycerol - for fluorescence microscopy is used as a mounting medium for stained histology specimens (e.g. toluidine blue stain, Cat. 104172 ). It is primarily designed to preserve the fluorescence of durable specimens for future diagnosis. 100 ml are sufficient for approx. 200 - 300 applications. The mounting agent is an IVD registered product and CE certified, thus can be used for clinical diagnostic purposes.
For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

  • Refractive index (20 °C) 1.472 - 1.474
  • Fluorescence (at 365 nm as quinine) < 100 ppb
  • Fluorescence (at 313 nm as quinine) < 100 ppb
  • Fluorescence (at 254 nm as quinine) < 100 ppb
  • Purity (20°C) > 99.5 %
  • Density 1.26 g/cm3

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service