1.04039
UVM II supplement
for Listeria spp., pkg of 1 vial, for the preparation of UVM-II Listeria Selective Enrichment Broth
Synonym(s):
UVM II supplement, Listeria Selective Supplement, Acriflavine hydrochloride, Panflavin
About This Item
Recommended Products
description
GranuCult™ Modified UVM broth (Base) (1.10824)
Quality Level
form
solid
autoignition temp.
>190 °C
potency
1048 mg/kg LD50, oral (Rat)
composition
Acriflavine hydrochloride, 13 mg per vial
packaging
pkg of 1 vial
technique(s)
microbiological culture: suitable
pH
1.0 (20 °C, 10 g/L in H2O)
mp
240-247 °C
solubility
300 g/L
bulk density
380 kg/m3
application(s)
food and beverages
microbiology
storage temp.
2-30°C
suitability
Listeria spp.
Related Categories
General description
Application
Analysis Note
Growth
- Listeria monocytogenes ATCC 19114 (WDCM 00110):
passes test
- Listeria ivanovii ATCC 19119 (WDCM 00018): passes test
- Staphylococcus aureus ATCC 6538-P (WDCM 00033): inhibited
Incubation: Listeria 24 hrs.; 30 °C; aerobic; other strains 24 hrs.; 35 °C; aerobic.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service