Skip to Content
Merck
All Photos(1)

Key Documents

1.02765

Supelco

Copper oxide

wire fine about 0.65 x 3 mm for elementary analysis (surface: CuO, core: Cu₂O) ACS

Synonym(s):

Copper oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-269-1
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

(oxygen demand as CuO): -≥ 55.0 %

form

solid

weight

=79.54 g/mol

mp

1300 °C

density

6.2 g/cm3 at 20 °C

bulk density

2200 kg/m3

cation traces

C: ≤0.002%
N: ≤0.002%

storage temp.

2-30°C

SMILES string

O=[Cu]

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Application


  • A study on a broadband photodetector based on hybrid 2D copper oxide/reduced graphene oxide.: This research developed a photodetector utilizing a novel hybrid of copper oxide and reduced graphene oxide, demonstrating enhanced broadband detection capabilities, which is significant for advancements in optical sensing technologies (Ngo et al., 2024).

  • Electro-sensing layer constructed of a WO(3)/CuO nanocomposite, for the electrochemical determination of 2-phenylphenol fungicide.: The paper discusses the fabrication and application of a tungsten oxide/copper oxide nanocomposite layer for sensitive and selective electrochemical detection of the fungicide 2-phenylphenol, highlighting its potential in environmental monitoring (Prabhu et al., 2023).

  • Copper Oxide Solution Sensor Formed on a Thin Film Having Nanowires for Detecting Ethanol in Water.: This study presents the development of a copper oxide-based sensor with nanowire-enhanced thin films for detecting ethanol in water, offering a promising approach for environmental and health-related applications (Kimura and Tohmyoh, 2022).

  • Tuning the Redox Chemistry of Copper Oxide Nanoarchitectures Integrated with rGOP via Facet Engineering: Sensing H(2)S toward SRB Detection.: This article explores the facet-engineered modification of copper oxide nanostructures combined with reduced graphene oxide, improving their redox properties for hydrogen sulfide sensing, which is crucial in detecting sulfate-reducing bacteria in industrial applications (Asif et al., 2022).

Analysis Note

Assay (oxygen demand as CuO): ≥ 55.0 %
Chloride (Cl): ≤ 0.005 %
Total carbon (C): ≤ 0.002 %
Total nitrogen (N): ≤ 0.002 %
Total sulfur (as SO₄): ≤ 0.005 %

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service