1.02407
di-Sodium oxalate
volumetric standard, secondary reference material for redox titration, traceable to NIST Standard Reference Material (SRM) Certipur® Reag. USP
Synonym(s):
di-Sodium oxalate, Oxalic acid sodium salt
About This Item
Recommended Products
Agency
ISO 17025
reag. USP
Quality Level
form
solid
potency
375 mg/kg LD50, oral (Rat)
technique(s)
titration: suitable
pH
8 (20 °C, 30 g/L in H2O)
mp
250-270 °C (decomposition)
density
2.27 g/cm3 at 20 °C
bulk density
600 kg/m3
storage temp.
15-25°C
SMILES string
[Na+].[Na+].[O-]C(=O)C(=O)[O-]
InChI
1S/C2H2O4.2Na/c3-1(4)2(5)6;;/h(H,3,4)(H,5,6);;/q;2*+1/p-2
InChI key
ZNCPFRVNHGOPAG-UHFFFAOYSA-L
General description
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Features and Benefits
- Available as a solid in a secure glass bottle to ensure its stability for the entire shelf life until opened.
- Certified reference material according to ISO 17034
- High-purity material traceable to NIST standard reference materials
- Qualified in a DIN EN ISO/IEC 17025 accredited laboratory
- In accordance with the chapter reagents of USP and Ph. Eur
- High-quality offering accurate titer determinations
- Accompanied by a certificate of analysis (CoA)
Packaging
Legal Information
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The titer determination or standardization of a titration solution is essential for accurate and reliable titration results.
The titer determination or standardization of a titration solution is essential for accurate and reliable titration results.
The titer determination or standardization of a titration solution is essential for accurate and reliable titration results.
The titer determination or standardization of a titration solution is essential for accurate and reliable titration results.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service