Skip to Content
Merck
All Photos(1)

Key Documents

1.02271

Supelco

Cerium(III) nitrate hexahydrate

extra pure

Synonym(s):

Cerium(III) nitrate hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ce(NO3)3 · 6 H2O
CAS Number:
Molecular Weight:
434.22
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-297-2
NACRES:
NA.21
Assay:
≥98.5% (complexometric)
form:
solid
solubility:
1754 g/L

Quality Level

Assay

≥98.5% (complexometric)

form

solid

potency

4200 mg/kg LD50, oral (Rat)

pH

3.7 (25 °C, 100 g/L in H2O)

solubility

1754 g/L

bulk density

1050 kg/m3

anion traces

chloride (Cl-): ≤0.003%

cation traces

Dy: ≤0.04%
Er: ≤0.2%
Eu: ≤0.04%
Fe: ≤0.001%
Ho: ≤0.02%
Nd: ≤0.02%
Pr: ≤0.01%
Sm: ≤0.02%
Tm: ≤0.02%
Y: ≤0.04%
heavy metals (as Pb): ≤0.002%

storage temp.

15-25°C

SMILES string

[Ce+3].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-]

InChI

1S/Ce.3NO3/c;3*2-1(3)4/q+3;3*-1

InChI key

HSJPMRKMPBAUAU-UHFFFAOYSA-N

Related Categories

Analysis Note

Assay (complexometric): ≥ 98.5 %
Chloride (Cl): ≤ 0.003 %
Heavy metals (as Pb): ≤ 0.002 %
Dy (Dysprosium): ≤ 0.04 %
Er (Erbium): ≤ 0.2 %
Eu (Europium): ≤ 0.04 %
Fe (Iron): ≤ 0.001 %
Ho (Holmium): ≤ 0.02 %
Nd (Neodymium): ≤ 0.02 %
Pr (Praseodym): ≤ 0.01 %
Sm (Samarium): ≤ 0.02 %
Tm (Thulium): ≤ 0.02 %
Y (Yttrium): ≤ 0.04 %

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service