1.01123
Withdrawal system for solvents
with manual pressue build-up, for use with 10 L and 30 L stainless steel barrels
Synonym(s):
Manual withdrawal system for barrels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121516
NACRES:
NA.03
Recommended Products
compatibility
for use with 10 L and 30 L stainless steel barrels
storage temp.
no temp limit
Related Categories
General description
Specially developed solvent withdrawal system with manual pressure build-up for EMD Millipore returnable stainless steel barrels of 10L and 30L. It includes two dip tubes, one for 10L and one for 30L. Specially made for maximum user safety and maximum application reliability.
with manual pressure build-up for 10 l and 30 l stainless steel barrels
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service