Skip to Content
Merck
All Photos(1)

Key Documents

1.01031

Supelco

Aluminium ammonium sulfate dodecahydrate

for analysis EMSURE® ACS

Synonym(s):

Aluminium ammonium sulfate dodecahydrate, Alum ammonium, Ammonium alum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Al(SO4)2 · 12 H2O
CAS Number:
Molecular Weight:
453.33
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-055-3
NACRES:
NA.21
Assay:
99.0-102.0% (complexometric)
grade:
ACS reagent
form:
solid

grade

ACS reagent

Quality Level

product line

EMSURE®

Assay

99.0-102.0% (complexometric)

form

solid

impurities

≤0.005% Insoluble matter

pH

2.6 (25 °C, 100 g/L in H2O)

bp

200 °C/1013 hPa

mp

93 °C

bulk density

700‑800 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
nitrate (NO3-): ≤0.001%

cation traces

As: ≤0.00005%
Ca: ≤0.05%
Cr: ≤0.0005%
Fe: ≤0.0002%
K: ≤0.005%
Mn: ≤0.0005%
Na: ≤0.005%
heavy metals (as Pb): ≤0.001%

storage temp.

2-30°C

SMILES string

[Al]1(O[S](=O)(=O)O1)O[S](=O)(=O)[O-].[N+H4].O.O.O.O.O.O.O.O.O.O.O.O

InChI

1S/Al.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

WZUKKIPWIPZMAS-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • The Individual Differences in the Perception of Oral Chemesthesis Are Linked to Taste Sensitivity.: Research highlights the link between chemesthetic perceptions and taste sensitivity, emphasizing the role of compounds like aluminum ammonium sulfate in sensory science, useful in tailoring consumer products to enhance palatability and efficacy (Sandell MA et al., 2021).

  • Compounding MgCl₂·6H₂O with NH₄Al(SO₄)₂·12H₂O or KAl(SO₄)₂·12H₂O to Obtain Binary Hydrated Salts as High-Performance Phase Change Materials.: Demonstrates the application of aluminum ammonium sulfate in creating efficient phase change materials for thermal energy storage, suggesting its broader applicability in sustainable energy solutions (Zhang Z et al., 2019).

  • Behavioral effects of long-term oral administration of aluminum ammonium sulfate in male and female C57BL/6J mice.: This animal study investigates the long-term effects of aluminum ammonium sulfate consumption, contributing to safety assessments and regulatory standards for its use in consumer products (Miyakawa T et al., 2018).

  • The Revision of Aluminum-containing Food Additive Provisions in China.: Discusses regulatory changes and safety evaluations of aluminum-based additives like aluminum ammonium sulfate in the food industry, impacting formulation and compliance strategies (Zhang JB et al., 2016).


Analysis Note

Assay (complexometric): 99.0 - 102.0 %
Insoluble matter: ≤ 0.005 %
Chloride (Cl): ≤ 0.0005 %
Nitrate (NO₃): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.001 %
As (Arsenic): ≤ 0.00005 %
Ca (Calcium): ≤ 0.05 %
Cr (Chromium): ≤ 0.0005 %
Fe (Iron): ≤ 0.0002 %
K (Potassium): ≤ 0.005 %
Mn (Manganese): ≤ 0.0005 %
Na (Sodium): ≤ 0.005 %
Corresponds to ACS

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service