1.00673
Phosphate Test, photometric
photometric, 3.0-100.0 mg/L (PO4-P), 9-307 mg/L (PO43-), 7-229 mg/L (P2O5), Spectroquant®
About This Item
Recommended Products
product name
Phosphate Cell Test, photometric, 3.0-100.0 mg/L (PO4-P), 9-307 mg/L (PO43-), 7-229 mg/L (P2O5), Spectroquant®
product line
Spectroquant®
Quality Level
usage
sufficient for 25 tests
specific analyte(s)
phosphate
measuring range
3.0-100.0 mg/L (PO4-P)
7-229 mg/L (P2O5)
9-307 mg/L (PO43-)
technique(s)
photometry: suitable
compatibility
for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600
detection method
photometric (Phosphomolybdenum blue)
storage temp.
15-25°C
1 of 4
This Item | 1.00474 | 1.00616 | 1.14543 |
---|---|---|---|
phosphate | phosphate | phosphate | phosphate |
photometry: suitable | photometry: suitable | photometry: suitable | photometry: suitable |
3.0-100.0 mg/L (PO4-P), 9-307 mg/L (PO43-), 7-229 mg/L (P2O5) | 0.05-5.00 mg/L (PO4-P), 0.11-11.46 mg/L (P2O5), 0.2-15.3 mg/L (PO43-) | 3.0-100.0 mg/L (PO4-P), 7-229 mg/L (P2O5), 9-307 mg/L (PO43-) | 0.05-5.00 mg/L (PO4-P), 0.11-11.46 mg/L (P2O5), 0.2-15.3 mg/L (PO43-) |
for use with Spectroquant® Move 100, for use with Spectroquant® Prove 100, for use with Spectroquant® Prove 600, for use with Spectroquant® Nova 60 A | for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100, for use with Spectroquant® Prove 300, for use with Spectroquant® Prove 600 | for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100, for use with Spectroquant® Prove 300, for use with Spectroquant® Prove 600 | for use with Spectroquant® Move 100, for use with Spectroquant® Nova 60 A, for use with Spectroquant® Prove 100, for use with Spectroquant® Prove 300, for use with Spectroquant® Prove 600 |
Spectroquant® | Spectroquant® | Spectroquant® | Spectroquant® |
General description
The method is analogous to EPA 365.2+3, APHA 4500-P E, and DIN EN ISO 6878.
The Spectroquant® Phosphate Cell Test allows the accurate quantification of phosphate. Elevated levels of phosphate might lead to eutrophication of water bodies, which is a risk to the environment. Therefore, the concentration of phosphate in drinking water and wastewater is regulated in many countries.
Further applications are the determination of phosphate in foods & beverages (e.g. beer) and in environmental and agricultural samples (e.g. soil).
The Spectroquant®Cell Tests come with prefilled 16 mm round cells and all the required reagents to perform the analysis according to the instruction leaflet provided.
All Spectroquant® Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode), which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service