Skip to Content
Merck
All Photos(2)

Key Documents

1.00327

Sigma-Aldrich

Hydrogen chloride solution

in ethanol, for microscopy

Synonym(s):

Hydrogen chloride – ethanol solution, Hydrogen chloride solution, Hydrochloric acid in ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCl · CH3CH2OH
CAS Number:
Molecular Weight:
36.46
Beilstein:
1098214
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

product name

Hydrogen chloride solution, in ethanol, for microscopy

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

pH

1.0 (20 °C in H2O)

transition temp

flash point 21 °C

density

0.90 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrochloric acid in ethanol - for microscopy, is used for human-medical cell diagnosis and serves the purpose of the bacteriological and histological investigation of sample material of human origin. Acid-fast bacteria are difficult to stain because of the high proportion of lipid and wax in their cell walls. The most efficient staining method is the hot staining according to Ziehl-Neelsen. In this method, carbolfuchsin solution is applied to the specimen and heated. This heating process accelerates the rate at which the fuchsin dye is absorbed and thus also that of the formation of the mycolatefuchsin complex in the cell wall. With the Kinyoun method, a modification of the AFB-Color carbol fuchsin solution renders heating unnecessary (cold staining). Once the acid-fast bacteria have absorbed the fuchsin dye, it is virtually impossible to decolorize them again, even when they are intensively treated with a decolorizing solution such as e. g. hydrochloric acid in ethanol (Product number 1.00327). Accordingly, acid-fast bacteria are termed as acid- and alcohol-fast for staining, and are stained red in the microscopic visualization. Correspondingly, all non-acid-fast microorganisms are counterstained with an appropriate dye. In this instruction for use malachite green is used for counterstaining.
Pretreatment of the specimens with Sputofluol dissolves the bacteria from the surrounding viscid sputum and cell material. It is an IVD and CE registered product and can be applied for diagnostical purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

For microscopy. Ziehl-Neelsen staining

Legal Information

SPUTOFLUOL is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service