Skip to Content
Merck
All Photos(1)

Key Documents

100065

Sigma-Aldrich

2-APB

≥97% (titration), crystalline solid, Ins(1,4,5)P3-induced Ca2+ release inhibitor, Calbiochem®

Synonym(s):

2-APB, 2-Aminoethoxydiphenylborate, (2-Aminoethoxy)diphenylborane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16BNO
CAS Number:
Molecular Weight:
225.09
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

2-APB, A cell-permeable modulator of Ins(1,4,5)P3-induced Ca2+ release.

Quality Level

description

RTECS - ED6150000

Assay

≥97% (titration)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 20 mg/mL
95% ethanol: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

B(OCCN)(c2ccccc2)c1ccccc1

InChI

1S/C14H16BNO/c16-11-12-17-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,11-12,16H2

InChI key

BLZVCIGGICSWIG-UHFFFAOYSA-N

General description

A cell-permeable modulator of Ins(1,4,5)P3-induced Ca2+ release. Inhibits Ins(1,4,5)P3-induced Ca2+ release from rat cerebellar microsomal preparations (IC50 = 42 µM) without affecting [3H]-Ins(1,4,5)P3 binding to its receptor. In liver cells, 2-APB inhibits store-operated Ca2+ channels through a mechanism which may involve its binding to either the channel protein or an associated regulatory protein. Has no effect on the Ca2+ release from the ryanodine-sensitive Ca2+ store prepared from rat leg skeletal muscle and heart.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Ins(1,4,5)P3-induced Ca2+ release
Product does not compete with ATP.
Reversible: no
Target IC50: 42 µM against Ins(1,4,5)P3-induced Ca2+ release from rat cerebellar microsomal preparations

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Brooke, R. T., et al. 2004. Lab. Invest.84, 29.
Gregory, R.B., et al. 2001. Biochem. J.354, 285.
Ma, H.T., et al. 2001. J. Biol. Chem.276, 18888.
Ascher-Landsberg, S., et al. 1999. Biochem. Biophys. Res. Commun.264, 979.
Maruyama, T., et al. 1997. J. Biochem.122, 498.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dominic D Quintana et al.
Journal of Alzheimer's disease : JAD, 75(1), 119-138 (2020-04-07)
Cerebrovascular pathology is pervasive in Alzheimer's disease (AD), yet it is unknown whether cerebrovascular dysfunction contributes to the progression or etiology of AD. In human subjects and in animal models of AD, cerebral hypoperfusion and hypometabolism are reported to manifest
Xuexin Zhang et al.
Cell calcium, 91, 102281-102281 (2020-09-09)
The ubiquitous Ca2+ release-activated Ca2+ (CRAC) channel is crucial to many physiological functions. Both gain and loss of CRAC function is linked to disease. While ORAI1 is a crucial subunit of CRAC channels, recent evidence suggests that ORAI2 and ORAI3
Bulat R Ramazanov et al.
Molecular biology of the cell, 35(4), ar50-ar50 (2024-01-31)
Ca2+ influx into the trans-Golgi Network (TGN) promotes secretory cargo sorting by the Ca2+-ATPase SPCA1 and the luminal Ca2+ binding protein Cab45. Cab45 oligomerizes upon local Ca2+ influx, and Cab45 oligomers sequester and separate soluble secretory cargo from the bulk
Kazuhiro Takano et al.
Development, growth & differentiation, 53(1), 37-47 (2011-01-26)
This study investigated the development of Ca²(+) signaling mechanisms and their role in initiating morphogenetic cell movement in the presumptive ectoderm of Japanese newt (Cynops pyrrhogaster) during gastrulation. Histochemical staining using fluorescently labeled ryanodine and dihydropyridine probes revealed that dihydropyridine
Prashanta Silwal et al.
Communications biology, 4(1), 548-548 (2021-05-12)
Mitochondrial function and innate immunity are intimately linked; however, the mechanisms how mitochondrion-shaping proteins regulate innate host defense remains largely unknown. Herein we show that mitofusin-2 (MFN2), a mitochondrial fusion protein, promotes innate host defense through the maintenance of aerobic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service