Skip to Content
Merck
All Photos(1)

Key Documents

09-433

Sigma-Aldrich

Anti-phospho-IRS1 (Tyr628) mouse/ (Tyr632) human Antibody

Upstate®, from rabbit

Synonym(s):

HIRS-1, Insulin Receptor Substrate 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr628/pTyr632 )

Gene Information

human ... IRS1(3667)

General description

Insulin receptor substrate (IRS) molecules are key mediators in insulin signaling and play a central role in maintaining basic cellular functions such as growth, survival, and metabolism. They act as docking proteins between the insulin receptor and a complex network of intracellular signaling molecules containing Src homology 2 (SH2) domains. Four members (IRS-1, IRS-2, IRS-3, IRS-4) of this family have been identified that differ as to tissue distribution, subcellular localization, developmental expression, binding to the insulin receptor, and interaction with SH2 domain-containing proteins. Results from targeted disruption of the IRS genes in mice have provided important clues to the functional differences among these related molecules, suggesting they play different and specific roles in vivo. The available data are consistent with the notion that IRS-1 and IRS-2 are not functionally interchangeable in tissues that are responsible for glucose production (liver), glucose uptake (skeletal muscle and adipose tissue), and insulin production (pancreatic β cells). In fact, IRS-1 appears to have its major role in skeletal muscle whereas IRS-2 appears to regulate hepatic insulin action as well as pancreatic β cell development and survival. By contrast, IRS-3 and IRS-4 genes appear to play a redundant role in the IRS signaling system. Defects in muscle IRS-1 expression and function have been reported in insulin-resistant states such as obesity and type 2 diabetes.

Specificity

Predicted to cross react with mouse and rat based on 100% sequence homology.
Recognizes phospho-IRS1 (Tyr628) mouse/ (Tyr632) human

Immunogen

peptide corresponding to amino acids encompassing and including phosphorylated Tyr628 in mouse and Tyr632 in human of IRS1

Application

Anti-phospho-IRS1 (Tyr628) mouse/ (Tyr632) human Antibody is an antibody against phospho-IRS1 (Tyr628) mouse/ (Tyr632) human for use in WB.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

routinely evaluated by western blot on lysates from untreated and insulin-treated (30 minutes) HEK 293 cells

Target description

170 kDa

Physical form

Affinity purified
Affinity purified rabbit polyclonal IgG in storage buffer containing PBS with 0.05% sodium azide in 50% glycerol.
Format: Purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanghee Park et al.
American journal of physiology. Endocrinology and metabolism, 326(3), E398-E406 (2024-02-07)
Resveratrol, a natural polyphenol compound contained in numerous plants, has been proposed as a treatment for obesity-related disease processes such as insulin resistance. However, in humans there are conflicting results concerning the efficacy of resveratrol in improving insulin action; the
Qutuba G Karwi et al.
Diabetes, obesity & metabolism, 21(8), 1944-1955 (2019-05-03)
Obesity is associated with high rates of cardiac fatty acid oxidation, low rates of glucose oxidation, cardiac hypertrophy and heart failure. Whether weight loss can lessen the severity of heart failure associated with obesity is not known. We therefore determined
Chao Liu et al.
Cancer medicine, 9(11), 3875-3884 (2020-04-06)
Several studies have suggested that drug resistance in colon cancer patients with diabetes may be associated with long-term insulin administration, which in turn decreases the survival rate. Metformin is a commonly used drug to treat diabetes but has been recently
Sanghee Park et al.
The Journal of physiology, 597(2), 449-466 (2018-11-11)
Exercise/exercise training can enhance insulin sensitivity through adaptations in skeletal muscle, the primary site of insulin-mediated glucose disposal; however, in humans the range of improvement can vary substantially. The purpose of this study was to determine if obesity influences the
Max Brown et al.
Molecular biology of the cell, 31(23), 2597-2629 (2020-09-03)
Accumulation of unfolded proteins in the endoplasmic reticulum (ER) causes ER stress and activates a signaling network known as the unfolded protein response (UPR). Here we characterize how ER stress and the UPR inhibit insulin signaling. We find that ER

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service