Recommended Products
biological source
rabbit
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
packaging
antibody small pack of 25 μg
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... AGO2(27161)
General description
This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing.
Provides endonuclease activity to RNA-induced silencing complexes (RISC). Cleaves siRNA/mRNA heteroduplexes bound to RISC. Essential for embryonic development as well as RNA-mediated gene silencing (RNAi).
Provides endonuclease activity to RNA-induced silencing complexes (RISC). Cleaves siRNA/mRNA heteroduplexes bound to RISC. Essential for embryonic development as well as RNA-mediated gene silencing (RNAi).
Specificity
Recognizes Ago 2, also known as eukaryotic translation initiation factor 2C, 2 (EIF2C 2), MGC3183 and Q10.
Immunogen
GST fusion protein corresponding to residues 7-48 of human argonaute 2 (Ago2).
Application
Anti-Ago 2 Antibody is a Rabbit Polyclonal Antibody for detection of Ago 2 also known as argonaute 2 & has been tested in WB.
Quality
Routinely evaluated by immunoblot in RIPA and cytosolic lysates from HeLa cells.
Target description
100 kDa
Physical form
Format: Purified
Analysis Note
Control
HeLa cell lysate, HL-60 cell lysate
HeLa cell lysate, HL-60 cell lysate
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Not finding the right product?
Try our Product Selector Tool.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nucleic acids research, 39(4), 1510-1525 (2010-10-26)
RNA interference is a powerful mechanism for sequence-specific inhibition of gene expression. It is widely known that small interfering RNAs (siRNAs) targeting the same region of a target-messenger RNA can have widely different efficacies. In efforts to better understand the
Identification of distinct miRNA target regulation between breast cancer molecular subtypes using AGO2-PAR-CLIP and patient datasets.
Genome Biology null
Limiting Ago protein restricts RNAi and microRNA biogenesis during early development in Xenopus laevis.
Genes & Development null
Molecular cancer, 23(1), 113-113 (2024-05-28)
The role of circRNAs in hepatocellular carcinoma (HCC) progression remains unclear. CircPIAS1 (circBase ID: hsa_circ_0007088) was identified as overexpressed in HCC cases through bioinformatics analysis. This study aimed to investigate the oncogenic properties and mechanisms of circPIAS1 in HCC development.
Cytogenetic and genome research, 162(1-2), 10-27 (2022-03-31)
Long noncoding RNA FOXD2 adjacent opposite strand RNA1 (FOXD2-AS1) plays an oncogenic role in various cancers, including gastric cancer (GC). However, the function of FOXD2-AS1 in regulating radiosensitivity of GC cells and its underlying molecular mechanisms have not been elucidated.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service