Skip to Content
Merck
All Photos(1)

Key Documents

07-585

Sigma-Aldrich

Anti-dimethyl-Histone H3 (Arg2) Antibody

serum, Upstate®

Synonym(s):

H3R2me2, Histone H3 (di methyl R2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, chicken

manufacturer/tradename

Upstate®

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

dimethylation (Arg2)

Gene Information

human ... H3F3B(3021)

Specificity

Broad species cross-reactivity expected
dimethyl-arginine at residue 2 of histone H3

Immunogen

peptide containing the sequence AdimRT, corresponding to dimethyl-arginine 2 of human histone H3

Application

Anti-dimethyl-Histone H3 (Arg2) Antibody is a Rabbit Polyclonal Antibody for detection of dimethyl-Histone H3 (Arg2) also known as H3R2me2, Histone H3 (di methyl R2) & has been validated in WB, PIA.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

routinely evaluated by immunoblot on acid-extracted proteins from chicken erythrocyte cells (Catalog # 13-107)

Target description

Mr 17kDa

Physical form

Serum
rabbit antiserum containing 0.05% sodium azide, before the addition of glycerol to 30%

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sinja Rakow et al.
Methods (San Diego, Calif.), 175, 53-65 (2019-09-23)
Among the widespread and increasing number of identified post-translational modifications (PTMs), arginine methylation is catalyzed by the protein arginine methyltransferases (PRMTs) and regulates fundamental processes in cells, such as gene regulation, RNA processing, translation, and signal transduction. As epigenetic regulators
Jihee Hong et al.
Journal of ginseng research, 46(3), 481-488 (2022-05-24)
Although the tumor-suppressive effects of ginsenosides in cell cycle have been well established, their pharmacological properties in mitosis have not been clarified yet. The chromosomal instability resulting from dysregulated mitotic processes is usually increased in cancer. In this study, we
PELP1 oncogenic functions involve alternative splicing via PRMT6.
Mann, M; Zou, Y; Chen, Y; Brann, D; Vadlamudi, R
Molecular Oncology null
Claudia Stein et al.
Nucleic acids research, 40(19), 9522-9533 (2012-08-21)
The protein arginine methyltransferase 6 (PRMT6) is a coregulator of gene expression and executes its repressing as well as activating function by asymmetric dimethylation of histone H3 at R2 (H3 R2me2a). Given that elevated expression levels of PRMT6 have been
Haolin Liu et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(37), E7717-E7726 (2017-08-30)
Two of the unsolved, important questions about epigenetics are: do histone arginine demethylases exist, and is the removal of histone tails by proteolysis a major epigenetic modification process? Here, we report that two orphan Jumonji C domain (JmjC)-containing proteins, JMJD5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service