Skip to Content
Merck
All Photos(1)

Key Documents

07-535

Sigma-Aldrich

Anti-MKP1 Antibody

Upstate®, from rabbit

Synonym(s):

Anti-CL100, Anti-HVH1, Anti-MKP-1, Anti-MKP1, Anti-PTPN10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... FARSB(10056)

Specificity

Recognizes MKP1.

Immunogen

peptide corresponding to amino acids 87-101 (DERSAALDGAKRDGT-C) of human MAP kinase phosphatase 1 (MKP1)

Application

Detect MKP1 using this Anti-MKP1 Antibody validated for use in WB.
Research Category
Signaling
Research Sub Category
Kinases & Phosphatases

Quality

routinely evaluated by immunoblot in RIPA lysates from 3T3/A31 cells

Target description

42 kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide as a preservative.
Protein A purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shen-Hsi Yang et al.
PLoS genetics, 8(12), e1003112-e1003112 (2012-12-29)
Embryonic stem cells and induced pluripotent stem cells represent potentially important therapeutic agents in regenerative medicine. Complex interlinked transcriptional and signaling networks control the fate of these cells towards maintenance of pluripotency or differentiation. In this study we have focused
Akira Imasato et al.
The Journal of biological chemistry, 277(49), 47444-47450 (2002-10-03)
Despite the importance of glucocorticoids in suppressing immune and inflammatory responses, their role in enhancing host immune and defense response against invading bacteria is poorly understood. We have demonstrated recently that glucocorticoids synergistically enhance nontypeable Haemophilus influenzae (NTHi)-induced expression of
Huali Sun et al.
Technology in cancer research & treatment, 22, 15330338231207765-15330338231207765 (2023-10-24)
Objectives: Dual specificity phosphatase 1 (DUSP1) is high-expressed in various cancers and plays an important role in the cellular response to agents that damage DNA. We aimed to investigate the expressions and mechanisms of DUSP1 signaling pathway regulating cytarabine (Ara-C)
C F Zheng et al.
The Journal of biological chemistry, 268(22), 16116-16119 (1993-08-05)
The activation of extracellular signal-regulated kinase (ERK) or mitogen-activated protein kinase (MAPK) by a dual specific kinase, MEK (MAPK or ERK kinase), is a critical event in the mitogenic signal transduction pathway. However, little is known about the mechanism of
Epinephrine and AICAR-induced PGC-1? mRNA expression is intact in skeletal muscle from rats fed a high-fat diet.
Frier, BC; Wan, Z; Williams, DB; Stefanson, AL; Wright, DC
American Journal of Physiology. Cell Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service