Skip to Content
Merck
All Photos(1)

Key Documents

07-476

Sigma-Aldrich

Anti-p75NTR (Neurotrophin Receptor) Antibody

Upstate®, from rabbit

Synonym(s):

CD271 antigen, Low affinity neurotrophin receptor p75NTR, Low-affinity nerve growth factor receptor, NGF receptor, low affinity nerve growth factor receptor, nerve growth factor receptor, nerve growth factor receptor (TNFR superfamily, member 16), p75 IC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, rat

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NGFR(4804)

General description

The Low Affinity Nerve Growth Factor Receptor (also called the LNGFR or p75 neurotrophin receptor) is one of the two receptor types for the neurotrophins, a family of protein growth factors which stimulate neuronal cells to survive and differentiate.
The neurotrophins are comprised of four proteins, all of which bind to the LNGFR: Nerve Growth Factor (NGF), Brain Derived Neurotrophic Factor (BDNF), Neurotrophin-3 (NT-3), and Neurotrophin-4 (NT-4).
Nerve growth factor, the prototypical growth factor, is a protein secreted by a neuron′s target. NGF is critical for the survival and maintenance of sympathetic and sensory neurons. NGF is released from the target cells, binds to and activates its high affinity receptor (TrkA), and is internalized into the responsive neuron. The NGF/TrkA complex is subsequently trafficked back to the cell body. This movement of NGF from axon tip to soma is thought to be involved in the long-distance signaling of neurons.

Specificity

Recognizes p75 neurotrophin receptor, MW ~75 kDa.

Immunogen

Epitope: IC domain
GST fusion protein corresponding to the intracellular domain (residues 274-425) of rat p75 neurotrophin receptor.

Application

Detect p75NTR (Neurotrophin Receptor) using this Anti-p75NTR (Neurotrophin Receptor) Antibody validated for use in IH, IP & WB.
Immunoprecipitation:
This antibody has been reported by an independent laboratory to immunoprecipitate p75 neurotrophin receptor.

Immunohistochemistry:
This antibody has been reported by an independent laboratory to detect p75 neurotrophin receptor in rat brain sections. (Roux, P., 1999)
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Quality

Evaluated by western blot in RIPA lysates from PC-12 cells.

Western Blot Analysis:
0.1-0.5 µg/mL of this antibody detected p75 neurotrophin receptor in RIPA lysates from PC-12 cells.

Target description

~75 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal IgG in buffer containing 70% storage buffer 0.1 M Tris-glycine, pH 7.4, 0.15 M NaCl, 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Brain tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypo-osmolar stress induces p75NTR expression by activating Sp1-dependent transcription.
Ramos, A; Ho, WC; Forte, S; Dickson, K; Boutilier, J; Favell, K; Barker, PA
The Journal of Neuroscience null
E W Neeley et al.
Neuroscience, 187, 24-35 (2011-04-19)
Psychiatric illness and anxiety disorders have strong neurodevelopmental components. Environmental insults such as prenatal exposure to stress and genetic differences in stress responses may affect brain development. A rat model of random variable prenatal stress was used to study the
Modulation of p75(NTR) prevents diabetes- and proNGF-induced retinal inflammation and blood-retina barrier breakdown in mice and rats.
Mysona, BA; Al-Gayyar, MM; Matragoon, S; Abdelsaid, MA; El-Azab, MF; Saragovi, HU; El-Remessy, AB
Diabetologia null
P P Roux et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 19(16), 6887-6896 (1999-08-06)
Seizure causes neuronal cell loss in both animal models and human epilepsy. To determine the contribution of apoptotic mechanisms to seizure-induced neuronal cell death, rat brains were examined for the occurrence of terminal deoxynucleotidyl transferase-mediated UTP nick end labeling (TUNEL)-positive
Deleted in breast cancer 1, a novel androgen receptor (AR) coactivator that promotes AR DNA-binding activity.
Fu, J; Jiang, J; Li, J; Wang, S; Shi, G; Feng, Q; White, E; Qin, J; Wong, J
The Journal of Biological Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service