Skip to Content
Merck
All Photos(1)

Key Documents

07-471

Sigma-Aldrich

Anti-Daxx Antibody

serum, Upstate®

Synonym(s):

Anti-BING2, Anti-DAP6, Anti-EAP1, Anti-SMIM40

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat, mouse, hamster

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

hamster ... Daxx(101836409)
human ... DAXX(1616)
mouse ... Daxx(13163)
rat ... Daxx(140926)

Specificity

Daxx

Immunogen

GST fusion protein corresponding to residues 472-740 of human Daxx.

Application

Anti-Daxx Antibody is an antibody against Daxx for use in IP, WB & IC.

Quality

routinely evaluated by immunoblot in HeLa nuclear extract

Target description

110 kDa

Linkage

Replaces: 04-445

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PML isoforms I and II participate in PML-dependent restriction of HSV-1 replication.
Cuchet, D; Sykes, A; Nicolas, A; Orr, A; Murray, J; Sirma, H; Heeren, J; Bartelt, A; Everett, RD
Journal of Cell Science null
Comparison of the biological and biochemical activities of several members of the alphaherpesvirus ICP0 family of proteins.
RD Everett, C Boutell, C McNair, L Grant, A Orr
Journal of virology null
Adenovirus type 5 early region 1B 55K oncoprotein-dependent degradation of cellular factor Daxx is required for efficient transformation of primary rodent cells.
Schreiner, S; Wimmer, P; Groitl, P; Chen, SY; Blanchette, P; Branton, PE; Dobner, T
Journal of virology null
Roger D Everett
Journal of virology, 90(1), 167-179 (2015-10-16)
Intrinsic immunity is an aspect of antiviral defense that operates through diverse mechanisms at the intracellular level through a wide range of constitutively expressed cellular proteins. In the case of herpesviruses, intrinsic resistance involves the repression of viral gene expression
Tetsuro Komatsu et al.
Journal of virology, 90(3), 1657-1667 (2015-11-27)
Promyelocytic leukemia protein nuclear bodies (PML-NBs) are subnuclear domains implicated in cellular antiviral responses. Despite the antiviral activity, several nuclear replicating DNA viruses use the domains as deposition sites for the incoming viral genomes and/or as sites for viral DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service