Skip to Content
Merck
All Photos(1)

Key Documents

07-315

Sigma-Aldrich

Anti-SGK1 Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, rat

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SGK1(6446)

Specificity

Recognizes SGK1.

Immunogen

peptide corresponding to amino acids 399-412 (C-GKSPDSVLVTASVK) of human SGK1

Application

Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling
This Anti-SGK1 Antibody is validated for use in IP, WB for the detection of SGK1.

Quality

routinely evaluated by immunoblot on RIPA lysates from A431 and Rat 2 cells

Target description

~50 kDa

Linkage

Replaces: 04-1027

Physical form

Format: Purified
Immunoaffinity Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide and 1.75mg/ml BSA before the addition of glycerol to 30%
Protein A purified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Rat 2 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucia D'Antona et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 35(5), 2006-2018 (2015-04-15)
Published observations on serum and glucocorticoid regulated kinase 1 (Sgk1) knockout murine models and Sgk1-specific RNA silencing in the RKO human colon carcinoma cell line point to this kinase as a central player in colon carcinogenesis and in resistance to
mTOR complex 2 (mTORC2) controls hydrophobic motif phosphorylation and activation of serum- and glucocorticoid-induced protein kinase 1 (SGK1).
Juan M Garcia-Martinez,Dario R Alessi
The Biochemical Journal null
C Filion et al.
The Journal of pathology, 217(1), 83-93 (2008-10-16)
The NR4A3 nuclear receptor is implicated in the development of extraskeletal myxoid chondrosarcoma (EMC), primitive sarcoma unrelated to conventional chondrosarcomas, through a specific fusion with EWSR1 resulting in an aberrant fusion protein that is thought to disrupt the transcriptional regulation
Acute activation of NHE3 by dexamethasone correlates with activation of SGK1 and requires a functional glucocorticoid receptor.
Wang, D; Zhang, H; Lang, F; Yun, CC
American Journal of Physiology. Cell Physiology null
Melissa L Vadnais et al.
Biology of reproduction, 89(5), 127-127 (2013-09-06)
Sperm motility encompasses a wide range of events involving epididymal maturation and activation of biochemical pathways, most notably cyclic AMP (cAMP)-protein kinase A (PKA) activation. Following the discovery of guanine-nucleotide exchange factors (RAPGEFs), also known as exchange proteins activated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service