06-996
Anti-phospho-Kip1 (Thr187) Antibody
Upstate®, from rabbit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, mouse, rat
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
phosphorylation (pThr187)
Gene Information
human ... CDKN1B(1027)
Specificity
phosphorylated Kip1
Immunogen
KLH-conjugated, synthetic peptide (C-PNAGSVEQ[pT]PKK) corresponding to amino acids 179-190 of human Kip1
Application
Anti-phospho-Kip1 (Thr187) Antibody is a high quality Rabbit Polyclonal Antibody for the detection of phospho-Kip1 (Thr187) & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Cell Cycle, DNA Replication & Repair
Quality
routinely evaluated by immunoblot on a HeLa cell RIPA lysate
Target description
27kDa
Physical form
0.2M Tris, 0.15 M NaCl, 0.1mM EDTA, 0.05% sodium azide before the addition of glycerol to 30%
Immunoaffinity Chromatography
Storage and Stability
1 year at -20°C
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Oncostatin M induces growth arrest by inhibition of Skp2, Cks1, and cyclin A expression and induced p21 expression.
Halfter, H; Friedrich, M; Resch, A; Kullmann, M; Stogbauer, F; Ringelstein, EB; Hengst, L
Cancer Research null
p27KIP1, an inhibitor of cyclin-dependent kinases.
Koff, A and Polyak, K
Progress in Cell Cycle Research, 1, 141-147 (1995)
G Wolf
Kidney international, 56(4), 1262-1265 (1999-10-03)
There are two fundamentally different growth responses for cells comprising the nephron: hyperplasia or hypertrophy. Cells that progress through the normal cell cycle double their DNA content and eventually divide during mitosis. Those cells that hypertrophy stop the growth process
J Kato
Frontiers in bioscience : a journal and virtual library, 4, D787-D792 (1999-11-30)
The first gap phase (G1) in the mammalian cell cycle plays a pivotal role in determining whether or not cells are to initiate DNA replication. Progression through G1 phase and transition into S phase are positively and negatively regulated by
Shuichan Xu et al.
The Journal of biological chemistry, 282(21), 15462-15470 (2007-04-06)
p27, an important cell cycle regulator, blocks the G(1)/S transition in cells by binding and inhibiting Cdk2/cyclin A and Cdk2/cyclin E complexes (Cdk2/E). Ubiquitination and subsequent degradation play a critical role in regulating the levels of p27 during cell cycle
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service