Skip to Content
Merck
All Photos(1)

Key Documents

06-758

Sigma-Aldrich

Anti-acetyl-p53 Antibody (Lys373, Lys382)

serum, Upstate®

Synonym(s):

Anti-BCC7, Anti-BMFS5, Anti-LFS1, Anti-P53, Anti-TRP53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

vertebrates

packaging

antibody small pack of 25 μL

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

acetylation (Lys373,Lys382)

Gene Information

human ... TP53(7157)

Related Categories

General description

p53 is a DNA-binding, oligomerization domain and transcription activation domain-containing tumor suppressor that upregulates growth arrest and apoptosis-related genes in response to stress signals, thereby influencing programmed cell death, cell differentiation and cell cycle control mechanisms. p53 localizes to the nucleus yet can be chaperoned to the cytoplasm by the negative regulator MDM2, an E3 ubiquitin ligase that is upregulated in the presence of active p53, where MDM2 polyubiquitinates p53 for proteasome targeting. p53 can assemble into tetramers in the absence of DNA, fluctuates between latent and active (DNA-binding) conformations, and is differentially activated through posttranslational modifications including phosphorylation and acetylation. Mutations in the DNA-binding domain (DBD) (amino acids 110-286) of p53 can compromise energetically favorable association with cis elements and are implicated in several human cancers.

Specificity

Recognizes p53 acetylated in vitro by recombinant p300, and for peptide containing acetyl-lysine 373 but not for the peptide containing acetyl-lysine 320.

Immunogen

peptide of human p53 corresponding to amino acids 368-389 with a duplication of amino acids 376 and 377 (HLKSKACKGQSTSTSRHKACKLMFKTC) where ACK denotes acetyl-lysine

Application

Detect acetyl-p53 (Lys373) with Anti-acetyl-p53 Antibody (Lys373, Lys382) (Rabbit Polyclonal Antibody), that has been shown to work in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Western Blot (on recombinant protein only)

Quality

routinely evaluated by an in vitro acetyl transferase assay on recombinant p300, HAT domain (Catalog #14-418)

Target description

53 kDa

Linkage

Replaces: 04-1137

Physical form

Rabbit polyclonal antiserum containing 0.05% sodium azide. Frozen solution.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Breast Cancer, Colon, Skin tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitative proteomic characterization of ethanol-responsive pathways in rat microglial cells.
Bell-Temin, Harris, et al.
Journal of Proteome Research, 12, 2067-2077 (2013)
Role for p300 in stabilization of p53 in the response to DNA damage.
Yuan, Z M, et al.
The Journal of Biological Chemistry, 274, 1883-1886 (1999)
Hye-Jung Park et al.
Cell journal, 21(1), 27-34 (2018-12-07)
Amentoflavone is the main component of Selaginella tamariscina widely known as an oriental traditional medicinal stuff that has been known to have a variety of medicinal effects such as the induction of apoptosis, antimetastasis, and anti-inflammation. However, the effect of
Circulatory miR34a as an RNAbased, noninvasive biomarker for brain aging.
Li, X; Khanna, A; Li, N; Wang, E
Aging null
Exercise Increases Markers of Spermatogenesis in Rats Selectively Bred for Low Running Capacity.
Torma, F; Koltai, E; Nagy, E; Ziaaldini, MM; Posa, A; Koch, LG; Britton, SL; Boldogh, I; Radak, Z
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service