Skip to Content
Merck
All Photos(3)

Key Documents

05-908

Sigma-Aldrich

Anti-RAD21 Antibody

Upstate®, from mouse

Synonym(s):

Anti-CDLS4, Anti-HR21, Anti-HRAD21, Anti-MCD1, Anti-MGS, Anti-NXP1, Anti-SCC1, Anti-hHR21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RAD21(5885)

General description

Aka Scc1, part of the cohesin complex, Rad21 is a nuclear protein that associates with mitotic chromatin and is essential for the association of kinetochores with microtubules. Rad21/Scc1 s is cleaved by caspases, promoting apoptosis.
The protein encoded by the Rad21 gene is highly similar to the gene product of Schizosaccharomyces pombe rad21, a gene involved in the repair of DNA double-strand breaks, as well as in chromatid cohesion during mitosis. This protein is a nuclear phospho-protein, which becomes hyperphosphorylated in cell cycle M phase.
RAD21 also plays a role in apoptosis, via its cleavage by caspase 3 or caspase 7 during early steps of apoptosis: the C-terminal 64 kDa cleavage product may act as a nuclear signal to initiate cytoplasmic events involved in the apoptotic pathway. Recent work has shown that the RAD21 has a role in the invasion and metastasis of cancer cells

Specificity

Rad21

Immunogen

GST fusion protein corresponding to residues 240-631 of human Rad21

Application

Detect RAD21 with Anti-RAD21 Antibody (Mouse Monoclonal Antibody), that has been shown to work in IP, WB, ICC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Quality

Routinely evaluated by immunoblot with HeLa nuclear extracts

Target description

100-120kDa

Physical form

Format: Purified
Protein G Purified
Prtotein G purified antibody in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CTCF controls expression and chromatin architecture of the human major histocompatibility complex class II locus.
Majumder, P; Boss, JM
Molecular and cellular biology null
Cohesin regulates MHC class II genes through interactions with MHC class II insulators.
Majumder, P; Boss, JM
Journal of immunology (Baltimore, Md. : 1950) (1950)
Peter G Wolf et al.
Journal of cell science, 131(11) (2018-05-05)
The DNA-embracing, ring-shaped multiprotein complex cohesin mediates sister chromatid cohesion and is stepwise displaced in mitosis by Wapl and separase (also known as ESPL1) to facilitate anaphase. Proper regulation of chromosome cohesion throughout meiosis is critical for preventing formation of
Scc1 sumoylation by Mms21 promotes sister chromatid recombination through counteracting Wapl.
Wu, N; Kong, X; Ji, Z; Zeng, W; Potts, PR; Yokomori, K; Yu, H
Genes & Development null
Johannes Buheitel et al.
The EMBO journal, 32(5), 666-676 (2013-01-31)
Faithful transmission of chromosomes during eukaryotic cell division requires sister chromatids to be paired from their generation in S phase until their separation in M phase. Cohesion is mediated by the cohesin complex, whose Smc1, Smc3 and Scc1 subunits form

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service