Skip to Content
Merck
All Photos(2)

Key Documents

05-182

Sigma-Aldrich

Anti-FAK Antibody, clone 2A7

clone 2A7, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A7, monoclonal

species reactivity

human, mouse, rat, avian

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTK2(5747)

Specificity

Recognizes FAK.

Immunogen

Mixture of affinity-purified tyrosine phosphoproteins from chick embryo fibroblasts expressing p125FAK.

Application

Anti-FAK Antibody, clone 2A7 detects level of FAK & has been published & validated for use in IC & IP.
Not recommended in Western Blot
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated by immunoprecipitation of FAK from a mouse 3T3/A31 RIPA cell lysate, subsequently immunobloted with Anti-FAK (Catalog #06-543)

Target description

125 kDa

Linkage

Replaces: 04-591

Physical form

Format: Purified
Protein G Purified
Protein G Purified immunoglobulin in Protein G Purified immunoglobulin in Purified immunoglobulin in 10mM PBS, pH 7.4 containing no preservatives.

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Expressed in most tissue, 3T3/A31 cell lysate

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Derkinderen et al.
Science (New York, N.Y.), 273(5282), 1719-1722 (1996-09-20)
Anandamide is an endogenous ligand for central cannabinoid receptors and is released after neuronal depolarization. Anandamide increased protein tyrosine phosphorylation in rat hippocampal slices and neurons in culture. The action of anandamide resulted from the inhibition of adenylyl cyclase and
Niki Tzenaki et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(12), 4840-4852 (2015-08-08)
The phosphatase and tensin homolog deleted on chromosome 10 (PTEN) tumor suppressor protein is regulated by various mechanisms that are not fully understood. This includes regulation by Tyr phosphorylation by a mechanism that remains elusive. Here, we show that focal
F Burgaya et al.
The Journal of biological chemistry, 272(45), 28720-28725 (1997-11-14)
pp125 focal adhesion kinase (FAK), a cytoplasmic tyrosine kinase transducing signals initiated by integrin engagement and G protein-coupled receptors, is highly expressed in brain. FAK from brain had a higher molecular weight and an increased autophosphorylation activity, than from other
Ambivalent role of pFAK-Y397 in serous ovarian cancer--a study of the OVCAD consortium.
Aust, S; Auer, K; Bachmayr-Heyda, A; Denkert, C; Sehouli, J; Braicu, I; Mahner et al.
Molecular Cancer null
Tyrosine phosphorylation of paxillin and focal adhesion kinase during insulin-like growth factor-I-stimulated lamellipodial advance.
Leventhal, P S, et al.
The Journal of Biological Chemistry, 272, 5214-5218 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service