Skip to Content
Merck
All Photos(5)

Key Documents

05-1075

Sigma-Aldrich

Anti-FOXO1 Antibody, clone 2H8.2

clone 2H8.2, from mouse

Synonym(s):

Forkhead box protein O1A, Forkhead in rhabdomyosarcoma, forkhead box O1, forkhead box O1A (rhabdomyosarcoma), forkhead homolog in rhabdomyosarcoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

2H8.2, monoclonal

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... FOXO1(2308)
mouse ... Foxo1(56458)

General description

Forkhead box O (FOXO) transcription factors play an important role in modulating metabolic functions. Given the relatively high expression of FOXO1 in insulin-responsive tissues, this transcription factor is highly poised to regulate energy metabolism. When nutrient and insulin levels are low, FOXO1 promotes expression of gluconeogenic enzymes. Conversely, in the fed state, insulin levels rise and stimulate uptake of glucose primarily into skeletal muscle and other organs, including adipose tissue. Under certain pathophysiologic conditions, including insulin resistance, negative signaling to FOXO1 is compromised. (Gross, D.N., 2009).

Specificity

This antibody detects FOXO1.
FOXO1 shuttles between the nucleus and the cytosol.

Immunogen

GST-tagged recombinant protein corresponding to full length FOXO1.

Application

Immunocytochemistry: A 1:500 dilution of a previous lot was used.

Immunocytochemistry: Representative lot data.
A 1:500 dilution of a previous lot was used.

Immunohistochemistry (Paraffin): Representative lot data.
Optimal Staining With Epitope Retrieval: Ductal Carcinoma In Situ.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-FOXO1 Antibody, clone 2H8.2 (Mouse Monoclonal Antibody) validated in WB, ICC, IHC(P) to detect FOXO1 also known as Forkhead box protein O1A, Forkhead in rhabdomyosarcoma.

Quality

Evaluated by western blot on HEK293 cell lysate.
Western Blot Analysis: 2 µg/mL of this antibody detected FOXO1 in HEK293 cell lysate.

Target description

~78-82 kDa

Linkage

Replaces: 04-1005

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
HEK293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianyu Pang et al.
International journal of molecular sciences, 24(17) (2023-09-09)
Transcription factors (TFs) have been shown to play a key role in the occurrence and development of tumors, including triple-negative breast cancer (TNBC), with a worse prognosis. Machine learning is widely used for establishing prediction models and screening key tumor
Dongjie Liu et al.
Oncology letters, 17(1), 450-455 (2019-01-19)
According to global cancer statistics in 2012, lung cancer (LC) was the most frequently diagnosed cancer and the leading cause of cancer-associated mortality among males worldwide. Owing to the limited therapeutic approaches available, novel methods for treating LC are required.
Jitendra Kumar et al.
Biological trace element research, 196(2), 552-564 (2019-12-13)
The excess fluoride intake has been shown to adversely affect male reproductive health. The aim of the present study was to investigate the key mechanism underlying fluoride-induced testicular dysfunction and the role of melatonin as a modulator of testicular metabolic
Francesca Luchetti et al.
Cells, 11(22) (2022-11-27)
Dysfunctional autophagy is linked to neuronal damage in ischemia/reperfusion injury. The Ras-related protein 7 (Rab7), a member of the Rab family of small GTPases, appears crucial for the progression of the autophagic flux, and its activity is strictly interconnected with
Jitendra Kumar et al.
Environmental toxicology, 36(3), 396-407 (2020-10-25)
BPS has detrimental effects on human reproductive health and emerged as an environmental contaminant for global health concern. This study deals with the adverse impact of BPS exposure on testicular oxidative stress, inflammation and apoptosis in adult male golden hamster

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service