Skip to Content
Merck
All Photos(1)

Key Documents

04-721

Sigma-Aldrich

Anti-Dicer1 Antibody, clone 5D12.2

clone 5D12.2, Upstate®, from mouse

Synonym(s):

DCR1, Dicer, HERNA, KIAA0928

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

5D12.2, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... DICER1(23405)

General description

RNA interference (RNAi) is mechanism for post-transcriptional gene silencing that leads to the loss of the activity of the gene. RNAi is activated when long dsRNAs are cleaved by the enzyme Dicer1 (RNase III family of dsRNA-specific endonucleases) into 21-23 nucleotide siRNA (small interfering RNA). These siRNAs bind proteins to form the RISC (RNA-induced silencing complex) which is activated when the double stranded siRNAs are unzipped to single stranded siRNAs. Activated RISC recognizes and binds complementary sequences in mRNA. The mRNA is cleaved and the overall effect is an inhibition of the protein expression and a loss of the gene activity. Dicer1 along with the Argonaute family of proteins (Ago1 and Ago2) share the same domain PIWI and PAZ domain that provide endonuclease activity to RNA-induced silencing complexes (RISC).

Specificity

Human Dicer1

Immunogen

KLH conjugated synthetic peptide corresponding to amino acids 378-385 of human Dicer1

Application

Anti-Dicer1 Antibody, clone 5D12.2 is a Mouse Monoclonal Antibody for detection of Dicer1 also known as DCR1, Dicer, HERNA, KIAA0928 & has been validated in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
RNA Metabolism & Binding Proteins

RNA Binding Protein (RBP)
Western blot

Quality

Routinely evaluated by western blot on lysates from HeLa nuclear extracts.

Target description

220 kDa

Physical form

Format: Purified
Protein G Chromatography

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Other Notes

Please refer to lot specific datasheet.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HongWei Liang et al.
Science China. Life sciences, 57(10), 973-981 (2014-10-01)
Ebola virus (EBOV), a member of the filovirus family, is an enveloped negative-sense RNA virus that causes lethal infections in humans and primates. Recently, more than 1000 people have been killed by the Ebola virus disease in Africa, yet no
Jiandong Shi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(10), 4381-4393 (2014-07-09)
MicroRNAs (miRNAs), including host miRNAs and viral miRNAs, play vital roles in regulating host-virus interactions. DNA viruses encode miRNAs that regulate the viral life cycle. However, it is generally believed that cytoplasmic RNA viruses do not encode miRNAs, owing to
Drosha mediates destabilization of Lin28 mRNA targets.
Qiao, C; Ma, J; Xu, J; Xie, M; Ma, W; Huang, Y
Cell Cycle null
Jiandong Shi et al.
Virology journal, 11, 121-121 (2014-07-02)
It is generally believed that RNA virus replicating in the cell cytoplasm would not encode microRNAs (miRNAs) due to nucleus inaccessibility. Recent studies have described cytoplasmic RNA virus genome-derived miRNAs in West Nile virus (WNV) and Dengue virus (DENV). However
Chatla Srinivas et al.
PloS one, 10(11), e0142006-e0142006 (2015-11-10)
Tumor microenvironment play role in angiogenesis and carcinogenesis. Etoposide, a known topoisomerase II inhibitor induces DNA damage resulting in cell cycle arrest. We developed a novel Etoposide analogue, Quinazolino-4β-amidopodophyllotoxin (C-10) that show better efficacy in regulating cell proliferation and angiogenesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service