04-085
Anti-Ago family Antibody
Upstate®, from mouse
Synonym(s):
argonaute family, piwi
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
monoclonal
species reactivity
human
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG1
NCBI accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... AGO1(26523)
Specificity
Ago family proteins
Immunogen
Synthetic peptide PKIDVYHYEVDIKPDKCPRRVNREVV
Application
Suggested dilution:
Western Blot: 1:500
Western Blot: 1:500
Research Category
Protein Trafficking
Protein Trafficking
Research Sub Category
RNA Metabolism & Binding Proteins
RNA Binding Protein (RBP)
RNA Metabolism & Binding Proteins
RNA Binding Protein (RBP)
Use Anti-Ago family Antibody (Mouse Monoclonal Antibody) validated in WB to detect Ago family also known as argonaute family PIWI.
Quality
Routinely evaluated by immunoblot.
Target description
~100 kDa
Physical form
100 μg of Protein A purified mouse monoclonal IgG1κ in 100μl of 1X PBS, pH 7.0, 0.1% Azide.
Format: Purified
Protein A purified
Storage and Stability
2 years at -20°C from date of shipment
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Analysis of the C. elegans Argonaute family reveals that distinct Argonautes act sequentially during RNAi
Cell, 127, 747-757 (2006)
PLoS genetics, 2(8), e134-e134 (2006-08-29)
Argonaute proteins are essential components of the molecular machinery that drives RNA silencing. In Drosophila, different members of the Argonaute family of proteins have been assigned to distinct RNA silencing pathways. While Ago1 is required for microRNA function, Ago2 is
Differential remodeling of mono- and trimethylated H3K27 during porcine embryo development.
Molecular Reproduction and Development null
Interactions between the RNA interference effector protein Ago1 and 14-3-3 proteins: consequences for cell cycle progression
The Journal of Biological Chemistry, 281, 37646-37651 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service