Skip to Content
Merck
All Photos(1)

Key Documents

Z-008

Supelco

Zolpidem-D7 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Zolpidem-D7 (Suitable for use with GC/MS or LC/MS) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14N3OD7
Molecular Weight:
314.43
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

100 μg/mL in methanol

technique(s)

GC/MS: suitable
LC/MS: suitable
gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[n]21c(nc(c2CC(=O)N(C)C)c3c(c(c(c(c3[2H])[2H])C([2H])([2H])[2H])[2H])[2H])C=CC(=C1)C

InChI

1S/C19H21N3O/c1-13-5-8-15(9-6-13)19-16(11-18(23)21(3)4)22-12-14(2)7-10-17(22)20-19/h5-10,12H,11H2,1-4H3/i1D3,5D,6D,8D,9D

InChI key

ZAFYATHCZYHLPB-UXSRMRDCSA-N

General description

Zolpidem, marketed as Ambien® or Stilnox®, is a nonbenzodiazepine used for the short-term treatment of insomnia. This stable-labeled internal standard is uniquely suited for GC/MS applications, because unlike the dimethyl-D6 analog, the deuterium label is retained in the major product ion fragment. As with dimethyl-D6 internal standard (Z-001), the D7-labeled analog is suitable for LC/MS methods.

Legal Information

Ambien is a registered trademark of Sanofi S.A.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Stilnox is a registered trademark of Sanofi S.A.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service