Skip to Content
Merck
All Photos(1)

Key Documents

T-905

Supelco

Thioridazine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2S2
CAS Number:
Molecular Weight:
370.57
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CSc1ccc2Sc3ccccc3N(CCC4CCCCN4C)c2c1

InChI

1S/C21H26N2S2/c1-22-13-6-5-7-16(22)12-14-23-18-8-3-4-9-20(18)25-21-11-10-17(24-2)15-19(21)23/h3-4,8-11,15-16H,5-7,12-14H2,1-2H3

InChI key

KLBQZWRITKRQQV-UHFFFAOYSA-N

General description

An analytical reference standard applicable for use as starting material in calibrators or controls for thioridazine testing methods by LC-MS/MS or GC-MS. Thioridazine is a treatment for schizophrenia sold under trade names Mellaril, Novoridazine, and Thioril.

Application


  • Fluorescence Chemosensor Development: Thioridazine solution has been utilized in the synthesis and application of an aldazine-based fluorescence chemosensor. This innovation allows for the sequential detection of Cu²⁺ and biological thiols in aqueous solutions and living cells, enhancing analytical capabilities in biochemical and medical research (Jia et al., 2016).

  • Voltammetric Analysis Enhancement: Research involving thioridazine solution has led to the development of a modified glassy carbon electrode with immobilized cobalt nanoparticles. This advancement facilitates the sensitive voltammetric determination of thioridazine, improving detection methods in pharmaceutical and clinical settings (Shahrokhian et al., 2009).

  • Microencapsulation Techniques: Thioridazine has been applied in pioneering work on the microencapsulation of drugs using gold nanoparticles. This technique aims to enhance drug delivery systems by improving the bioavailability and controlled release of antipsychotic medications, offering significant benefits for long-term psychiatric treatment (Lai et al., 2006).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

It is anticipated that as is usually the case, financial motivation will move TZ from its current status “potential anti-MDR/XDR agent,” to one that will cure MDR and XDR TB. Foreword.
Leonard Amaral
Recent patents on anti-infective drug discovery, 6(2), 76-76 (2011-07-30)
Diana Machado et al.
PloS one, 7(4), e34538-e34538 (2012-04-12)
Multidrug resistant (MDR) tuberculosis is caused by Mycobacterium tuberculosis resistant to isoniazid and rifampicin, the two most effective drugs used in tuberculosis therapy. Here, we investigated the mechanism by which resistance towards isoniazid develops and how overexpression of efflux pumps
Maija Purhonen et al.
Pharmacoepidemiology and drug safety, 21(11), 1227-1231 (2012-09-04)
Thioridazine is a first-generation antipsychotic drug that was withdrawn from the market worldwide in 2005. The outcome of clinically stable schizophrenia patients who used thioridazine before market withdrawal was evaluated. Nationwide registers in Finland were utilized to study thioridazine use
Eduardo Abbate et al.
The Journal of antimicrobial chemotherapy, 67(2), 473-477 (2011-12-03)
Current drug choices to treat extensively drug-resistant (XDR) tuberculosis (TB) are scarce; therefore, information on the safety, tolerability and efficacy of alternative regimens is of utmost importance. The aim of this study was to describe the management, drug adverse effects
Gabriella Spengler et al.
International journal of antimicrobial agents, 39(1), 16-21 (2011-10-11)
Thioridazine (TZ)-induced accumulation of the universal efflux pump substrate ethidium bromide and its subsequent efflux by Salmonella strains with various degrees of overexpressed efflux pumps takes place automatically at pH 7.4, is independent of a metabolic source, is not affected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service