Skip to Content
Merck
All Photos(1)

Key Documents

T-043

Supelco

Trenbolone solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22O2
CAS Number:
Molecular Weight:
270.37
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

C[C@]12C=CC3=C4CCC(=O)C=C4CCC3C1CC[C@@H]2O

InChI

1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h8-10,15-17,20H,2-7H2,1H3/t15?,16?,17-,18-/m0/s1

InChI key

MEHHPFQKXOUFFV-FOIPXRHGSA-N

General description

Trenbolone is a veterinary steroid used to increase muscle growth in livestock and has been used illicitly by athletes as a performance-enhancing drug. This Snap-N-Spike® reference solution is suitable for use in LC/MS or GC/MS applications in sports testing, clinical toxicology, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D R Ekman et al.
Environmental science & technology, 46(17), 9673-9680 (2012-08-01)
Widespread environmental contamination by bisphenol A (BPA) has created the need to fully define its potential toxic mechanisms of action (MOA) to properly assess human health and ecological risks from exposure. Although long recognized as an estrogen receptor (ER) agonist
Erica K Brockmeier et al.
Aquatic toxicology (Amsterdam, Netherlands), 128-129, 163-170 (2013-01-15)
The Eastern and Western mosquitofish (Gambusia holbrooki and G. affinis) are potential bioindicator organisms for endocrine disruptors. Male mosquitofish have an elongated anal fin (gonopodium) used for internal fertilization whose formation is driven by androgens. Normal female mosquitofish have a
B R Blackwell et al.
Ecotoxicology and environmental safety, 85, 110-114 (2012-09-07)
Manure from livestock feeding operations is commonly applied to agricultural fields as an alternative to commercial fertilizers. Trenbolone acetate (TbA) is a synthetic growth promoter frequently utilized in beef cattle feeding operations. Metabolites of TbA can be present in manure
Bushra Khan et al.
Chemosphere, 89(11), 1443-1449 (2012-07-17)
The increasing size of concentrated animal feeding operations has led to a concomitant increase in the land-application of manure, which has spawned research on the concentrations and environmental risk assessment of natural and synthetic hormones in animal manures. 17β-Trenbolone acetate
Allen W Olmstead et al.
Environmental toxicology and chemistry, 31(10), 2391-2398 (2012-08-01)
Trenbolone is an androgen agonist used in cattle production and has been measured in aquatic systems associated with concentrated animal-feeding operations. In this study, the authors characterized the effects of aqueous exposure to 17β-trenbolone during larval Xenopus tropicalis development. Trenbolone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service