Skip to Content
Merck
All Photos(1)

Key Documents

S-056

Supelco

JWH-122 4-Hydroxypentyl metabolite-D5 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20D5NO2
Molecular Weight:
376.50
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

Quality Level

grade

certified reference material

form

liquid

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=C([2H])N(CCCC(C)O)C2=C1C([2H])=C([2H])C([2H])=C2[2H])C3=CC=C(C)C4=C3C=CC=C4

InChI

1S/C25H25NO2/c1-17-13-14-22(20-10-4-3-9-19(17)20)25(28)23-16-26(15-7-8-18(2)27)24-12-6-5-11-21(23)24/h3-6,9-14,16,18,27H,7-8,15H2,1-2H3/i5D,6D,11D,12D,16D

InChI key

CLPJTHSXFZPIBP-DHJLQICESA-N

General description

An internal standard suitable for quantitation of JWH-122 4-Hydroxypentyl metabolite in biological matrices such as urine for LC/MS or GC/MS testing applications in clinical toxicology, forensic analysis, or urine drug testing. This monohydroxy analog is a primary urinary metabolite of JWH-122, a synthetic cannabinoid found in several blends of the herbal mixture Spice. The biomarker is also referred to as the omega-1 hydroxy metabolite of JWH-122.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service