Skip to Content
Merck
All Photos(1)

Key Documents

O-901

Supelco

Oxazepam-d5 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D5H6ClN2O2
CAS Number:
Molecular Weight:
291.74
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

shipped in

wet ice

storage temp.

−20°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C2=NC(O)C(=O)Nc3ccc(Cl)cc23

InChI

1S/C15H11ClN2O2/c16-10-6-7-12-11(8-10)13(9-4-2-1-3-5-9)18-15(20)14(19)17-12/h1-8,15,20H,(H,17,19)/i1D,2D,3D,4D,5D

InChI key

ADIMAYPTOBDMTL-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxazepam is a benzodiazepine drug used to treat anxiety and insomnia and to control the symptoms of alcohol withdrawal. This stable-labeled internal standard is suitable for isotope dilution methods or the quantitation of oxazepam levels using GC/MS or LC/MS in testing applications such as clinical toxicology, urine drug testing, forensic analysis or pain prescription monitoring.

Application

  • Application as an internal standard: Oxazepam-d5 serves as an internal standard for the gas chromatography-mass spectrometry analysis of benzodiazepines, enhancing accuracy and precision in quantifying drugs like lorazepam in biological samples (Pichini et al., 1999).
  • Quantification of benzodiazepine metabolites: Utilized in the sensitive detection and quantification of flunitrazepam metabolites in urine, demonstrating its role in forensic toxicology to identify drug use patterns (elSohly et al., 1997).
  • Enhancement of mass spectrometry methodologies: Oxazepam-d5 aids in improving the methods of mass spectrometry by serving as a standard reference material, crucial for the precise measurement of benzodiazepine metabolites (Meatherall, 1994).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niclas Stephanson et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 985, 189-196 (2015-02-18)
A mass spectrometric method for drugs of abuse testing in exhaled breath employing a sampling device collecting aerosol particles was developed and applied in routine use. Analytes covered were amphetamine, methamphetamine, 6-acetylmorphine, morphine, cocaine, benzoylecgonine, diazepam, oxazepam and tetrahydrocannabinol. The
Bernhard Watzer et al.
International journal of legal medicine, 129(4), 759-769 (2014-12-20)
Arthrotec(®) (AT) is a combination of diclofenac, a nonsteroidal anti-inflammatory drug (NSAID), and misoprostol (MP), a synthetic analogue of prostaglandin E1 (PGE1). MP is a lipophilic methyl ester prodrug. It is readily metabolized to the biologically active misoprostol acid (MPA).
André Valle de Bairros et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 975, 24-33 (2014-12-03)
In this study, it is shown a method for the determination of benzodiazepines and their main metabolites in urine samples by hollow-fiber liquid-phase microextraction (LPME) in the three-phase mode. Initially, the hydrolysis step was performed using 100 μL of sodium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service