Skip to Content
Merck
All Photos(1)

Key Documents

N-051

Supelco

Nalbuphine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28ClNO4
CAS Number:
Molecular Weight:
393.90
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.O[C@H]1CC[C@@]2(O)[C@H]3Cc4ccc(O)c5O[C@@H]1[C@]2(CCN3CC6CCC6)c45

InChI

1S/C21H27NO4.ClH/c23-14-5-4-13-10-16-21(25)7-6-15(24)19-20(21,17(13)18(14)26-19)8-9-22(16)11-12-2-1-3-12;/h4-5,12,15-16,19,23-25H,1-3,6-11H2;1H/t15-,16+,19-,20-,21+;/m0./s1

InChI key

YZLZPSJXMWGIFH-BCXQGASESA-N

Gene Information

General description

A Snap-N-Spike® reference solution applicable for use in urine drug testing, pain prescription monitoring, clinical toxicology, forensic analysis, or pharmaceutical research by LC-MS/MS or GC/MS. Nalbuphine is a semisynthetic opioid analgesic sold under the trade name Nubain®. This painkiller is prescribed for the relief of moderate to severe pain.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Nubain is a registered trademark of Endo Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David Sanchez-Migallon Guzman et al.
American journal of veterinary research, 72(6), 736-740 (2011-06-02)
To evaluate the antinociceptive effects and duration of action of nalbuphine HCl administered IM on thermal thresholds in Hispaniolan Amazon parrots (Amazona ventralis). 14 healthy adult Hispaniolan Amazon parrots of unknown sex. 3 doses of nalbuphine (12.5, 25, and 50
Robert Gear et al.
PloS one, 8(1), e50169-e50169 (2013-01-24)
Nalbuphine, an agonist-antagonist kappa-opioid, produces brief analgesia followed by enhanced pain/hyperalgesia in male postsurgical patients. However, it produces profound analgesia without pain enhancement when co-administration with low dose naloxone. To examine the effect of nalbuphine or nalbuphine plus naloxone on
Jason S Clark et al.
Veterinary ophthalmology, 14(6), 358-364 (2011-11-05)
To evaluate the effectiveness of topical nalbuphine or oral tramadol in the treatment of corneal pain in dogs. Fourteen male Beagle dogs. Dogs were divided into three treatment groups and sedated with dexmedetomidine (5 μ/kg IV). A 4 mm corneal
Juan F De La Mora-González et al.
Middle East journal of anaesthesiology, 21(4), 553-557 (2013-01-19)
Dexmedetomidine has demonstrated to be useful in several clinical fields due to its respiratory safety and cardiovascular stability. We undertook this study to determine its usefulness in plastic surgery. Sixty patients were divided into two parallel groups. A group received
S Sultana et al.
Drug research, 63(5), 224-227 (2013-03-15)
The interaction of nalbuphine hydrochloride with calf thymus deoxyribonucleic acid was investigated by absorption and fluorescence titration techniques. Hypochromic effect was observed in the absorption spectra of nalbuphine. The fluorescence quenching of nalbuphine by DNA was found to be static

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service