Skip to Content
Merck
All Photos(1)

Key Documents

M-096

Supelco

Morphine-6-β-D-glucuronide solution

100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27NO9
CAS Number:
Molecular Weight:
461.46
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol: water (1:1)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CN1CC[C@]23[C@H]4Oc5c(O)ccc(C[C@@H]1[C@@H]2C=C[C@@H]4O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(O)=O)c35

InChI

1S/C23H27NO9/c1-24-7-6-23-10-3-5-13(31-22-17(28)15(26)16(27)19(33-22)21(29)30)20(23)32-18-12(25)4-2-9(14(18)23)8-11(10)24/h2-5,10-11,13,15-17,19-20,22,25-28H,6-8H2,1H3,(H,29,30)/t10-,11+,13-,15-,16-,17+,19-,20-,22+,23-/m0/s1

InChI key

GNJCUHZOSOYIEC-GAROZEBRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Morphine-6-β-D-glucuronide is a primary plasma and urinary metabolite of the opiate analgesic morphine. Sold under trade names including MS Contin®, Kadian®, Oramorph®, and Roxanol for pain relief, morphine is also a commonly abused recreational drug. This certified analytical reference standard is suitable for a variety of GC/MS or LC/MS applications from pain prescription monitoring and urine drug testing to clinical toxicology and forensic analysis.

Application

  • Pharmacokinetics and Clinical Study Applications: A method involving liquid chromatography coupled to tandem mass spectrometry was developed and validated for the simultaneous quantification of several analgesics, including Morphine-6-β-D-glucuronide, applied in a clinical study for determining neurological death in intensive care units (Jutras et al., 2020).
  • Opioid Receptor Research: Research highlighted the higher efficacy of Morphine-6-β-D-glucuronide compared to morphine as a mu-opioid receptor agonist in the rat locus coeruleus, supporting its use in detailed opioid receptor studies (Osborne et al., 2000).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Kadian is a registered trademark of Actavis Elizabeth LLC
MS Contin is a registered trademark of Purdue Frederick Company
Oramorph is a registered trademark of AAIPharma, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

75.2 °F - closed cup

Flash Point(C)

24 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claire Trécant et al.
European journal of medicinal chemistry, 46(9), 4035-4041 (2011-06-22)
Synthesis and biological evaluation of new derivatives of Morphine-6-Glucuronide (M6G) are described. M6G is an active metabolite of morphine which displays more analgesia than morphine with a superior side effect profile but with a less efficiently BBB penetration. These phenomena
J A Hannam et al.
Anaesthesia and intensive care, 40(5), 867-870 (2012-09-01)
A morphine plasma concentration/respiratory rate relationship has been described for both adults and children although that of its metabolite, morphine-6-glucuronide, remains uncertain. We describe this relationship in a child with end-stage renal failure who received repeat morphine administration over two
Claudia F Clavijo et al.
Analytical and bioanalytical chemistry, 400(3), 715-728 (2011-03-15)
Opioids such as morphine are the cornerstone of pain treatment. The challenge of measuring the concentrations of morphine and its active metabolites in order to assess human pharmacokinetics and monitor therapeutic drugs in children requires assays with high sensitivity in
Toyofumi Suzuki
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 131(10), 1445-1451 (2011-10-04)
Opioid analgesics exhibit cationic properties under physiological conditions, and the mechanism underlying permeation of the blood-brain barrier thus cannot be fully explained by simple diffusion alone. Various types of transporters that exhibit substrate specificity are localized on the blood-brain barrier
Senthilkumar Sadhasivam et al.
Journal of opioid management, 8(4), 217-226 (2012-09-04)
Interindividual variability in analgesic response and adverse effects of opioids because of narrow therapeutic indices are major clinical problems. Morphine is an opioid commonly used in children to manage perioperative pain. Al-though size and age often are considered primary covariates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service