Skip to Content
Merck
All Photos(1)

Key Documents

M-077

Supelco

1-Mononitroglycerin solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO5
CAS Number:
Molecular Weight:
137.09
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

O[C@H](CO)CO[N+]([O-])=O

InChI

1S/C3H7NO5/c5-1-3(6)2-9-4(7)8/h3,5-6H,1-2H2

InChI key

HXWLJBVVXXBZCM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Mononitroglycerin is a major plasma metabolite of the pharmaceutical vasodilator nitroglycerin. This Certified Spiking Solution® is suitable for use in chromatography or mass spectrometry-based applications in pharmaceutical research or clinical toxicology. Nitroglycerin is used to treat heart conditions such as angina and chronic heart failure.

Application


  • Growth of Arthrobacter sp. strain JBH1 on Nitroglycerin: This study highlights the novel capability of Arthrobacter sp. strain JBH1 to metabolize nitroglycerin as its sole source of carbon and nitrogen. The findings are critical for biotechnological applications aiming to biodegrade nitroglycerin residues in contaminated environments, presenting an effective bioremediation strategy for pollutants derived from pharmaceutical manufacturing processes (Husserl et al., 2010).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Zimmermann et al.
Arzneimittel-Forschung, 44(4), 474-477 (1994-04-01)
The enantiomers of glyceryl-1-nitrate, a metabolite of glyceryl trinitrate, were pharmacologically characterized in vitro and in animals. In the Langendorff heart (l) G-1-N was double as potent as (d) G-1-N with respect to the enhancement of coronary flow. The two
M Leitold et al.
Arzneimittel-Forschung, 36(5), 814-821 (1986-05-01)
The present studies yield that all 4 metabolites of glyceryl trinitrate (Nitro Mack, GTN) cause the same typical pharmacological effects as the parent substance. Continuous infusion of 4 mg/kg/min of glyceryl 1-nitrate (G-1-N) in the conscious dog results in a
H Laufen et al.
European journal of clinical pharmacology, 31(2), 169-175 (1986-01-01)
The plasma kinetics and urinary excretion of glycerol-1-nitrate (G-1-N), a water soluble metabolite of glycerol trinitrate with anti-anginal potential, have been investigated in healthy human volunteers following oral doses of 10, 20 and 40 mg tablets and 20 mg as
H Laufen et al.
British journal of clinical pharmacology, 23(3), 287-293 (1987-03-01)
The plasma kinetics and urinary excretion of glyceryl-1-nitrate (G-1-N), a metabolite of glyceryl trinitrate with antianginal potential, were investigated in 10 healthy male volunteers, after intravenous infusion and oral administration of 20 mg G-1-N. The apparent volume of G-1-N distribution
C Romanin et al.
Journal of molecular and cellular cardiology, 20(5), 389-396 (1988-05-01)
Nitrovasodilators relax vascular smooth muscle by stimulating guanylate cyclase. Ignarro et al. (1981) proposed a mechanistic scheme according to which organic nitrates release nitrite in the presence of thiols. The corresponding nitrous acid would decay leading to nitric oxide, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service