Skip to Content
Merck
All Photos(1)

Key Documents

M-008

Supelco

(±)-Methadone-D3 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

DL-Methadone-d3 solution, DL-6-Dimethylamino-4,4-diphenyl-3-heptanone-1,1,1-d3 solution, DL-Methadone-(heptyl-1,1,1-d3) solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D3H24NO
CAS Number:
Molecular Weight:
312.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

mass shift

M+3

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])CC(=O)C(CC(C)N(C)C)(c1ccccc1)c2ccccc2

InChI

1S/C21H27NO/c1-5-20(23)21(16-17(2)22(3)4,18-12-8-6-9-13-18)19-14-10-7-11-15-19/h6-15,17H,5,16H2,1-4H3/i1D3

InChI key

USSIQXCVUWKGNF-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A stable-labeled internal standard applicable for use in isotope dilution methods or quantitation of methadone levels by GC/MS or LC/MS in forensic analysis, urine drug testing, pain prescription monitoring or clinical toxicology applications. Methadone is a synthetic opioid used medically as an analgesic and as a maintenance anti-addictive for use in patients with opioid dependency. The drug is sold under several trade names such as Symoron, Dolophine®, Amidone, Methadose®, Physeptone, or Heptadon.

Application


  • Forensic Toxicology Applications: Methadone-D3 is utilized as an internal standard in liquid chromatography-tandem mass spectrometry (LC-MS/MS) for the quantification of methadone-D3 in biological specimens. These isotopic labels are critical in forensic toxicology for the precise quantification of drugs and their metabolites in biological specimens, thereby enhancing the accuracy of drug monitoring in forensic cases (Johansen and Jensen, 2004).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Dolophine is a registered trademark of Roxane Laboratories, Inc.
Methadose is a registered trademark of Mallinckrodt Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service