I-026
Interference Mix 4 (For Qualitative Use Only)
5-100 μg/mL each component in methanol, Cerilliant®
Synonym(s):
Qualitative Interference Mixture
About This Item
Recommended Products
grade
analytical standard
Quality Level
form
liquid
composition
Alprazolam, 100 μg/mL
Cimetidine, 100 μg/mL
Citalopram hydrobromide, 100 μg/mL (as free base)
Clonazepam, 100 μg/mL
Clopidogrel bisulfate, 100 μg/mL (as free base)
Dextromethorphan, 50 μg/mL
Fluconazole, 100 μg/mL
Hydrochlorothiazide, 100 μg/mL
Lamotrigine, 100 μg/mL
Methylphenidate hydrochloride, 100 μg/mL (as free base)
Omeprazole, 100 μg/mL
L-Thyroxine, 5 μg/mL
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
psicótropo (Spain); Decreto Lei 15/93: Tabela IV/IIB (Portugal)
concentration
5-100 μg/mL each component in methanol
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
49.5 °F - closed cup
Flash Point(C)
9.7 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service