Skip to Content
Merck
All Photos(1)

Key Documents

H-125

Supelco

5beta/6beta-Hydroxy Lurasidone (Mixture of Diastereomers) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H36N4O3S
Molecular Weight:
508.68
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

General description

Hydroxylurasidone is the most abundant metabolite of lurasidone in urine. Lurasidone, marketed under the trade name Latuda®, is a new atypical antipsychotic developed for the treatment of schizophrenia and episodes of depression in adults with bipolar disorder (manic depression). A certified solution standard suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, pharmaceutical research, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Latuda is a registered trademark of Dainippon Sumitomo Pharma Co., Ltd.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina J Matheny et al.
Chemistry & biology, 20(11), 1352-1363 (2013-11-05)
Phenotypic high-throughput chemical screens allow for discovery of small molecules that modulate complex phenotypes and provide lead compounds for novel therapies; however, identification of the mechanistically relevant targets remains a major experimental challenge. We report the application of sequential unbiased
Eleonora Lapi et al.
Molecular cell, 32(6), 803-814 (2008-12-30)
p73 has been identified as a structural and functional homolog of the tumor suppressor p53. The transcriptional coactivator Yes-associated protein (YAP) has been demonstrated to interact with and to enhance p73-dependent apoptosis in response to DNA damage. Here, we show
Kenneth D Harrison et al.
The EMBO journal, 30(12), 2490-2500 (2011-05-17)
Dolichol monophosphate (Dol-P) functions as an obligate glycosyl carrier lipid in protein glycosylation reactions. Dol-P is synthesized by the successive condensation of isopentenyl diphosphate (IPP), with farnesyl diphosphate catalysed by a cis-isoprenyltransferase (cis-IPTase) activity. Despite the recognition of cis-IPTase activity
Peng Wang et al.
Nature medicine, 21(4), 383-388 (2015-03-10)
Types 1 and 2 diabetes affect some 380 million people worldwide. Both ultimately result from a deficiency of functional pancreatic insulin-producing beta cells. Beta cells proliferate in humans during a brief temporal window beginning around the time of birth, with
Cristin D Davidson et al.
Annals of clinical and translational neurology, 3(5), 366-380 (2016-05-28)
Niemann-Pick type C (NPC) disease is a fatal, neurodegenerative, lysosomal storage disorder characterized by intracellular accumulation of unesterified cholesterol (UC) and other lipids. While its mechanism of action remains unresolved, administration of 2-hydroxypropyl-β-cyclodextrin (HPβCD) has provided the greatest disease amelioration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service