Skip to Content
Merck
All Photos(1)

Key Documents

F-018

Supelco

4-Fluoroamphetamine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12FN · HCl
CAS Number:
Molecular Weight:
189.66
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

NC(C)CC1=CC=C(F)C=C1.Cl

InChI

1S/C9H12FN.ClH/c1-7(11)6-8-2-4-9(10)5-3-8;/h2-5,7H,6,11H2,1H3;1H

InChI key

GKWYMWZWSCKSMT-UHFFFAOYSA-N

General description

This Certified Spiking Solution® is suitable for numerous applications in GC/MS or LC/MS testing of 4-fluoroamphetamine from clinical toxicology and urine drug testing to forensic analysis. A fluoro analog of amphetamine, 4-Fluoroamphetamine is sold in the illicit market as a club/designer drug under street names including R2D2, Flux and Flits. The drug produces stimulant and possibly empathogenic or psychoactive effects.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F McElroy et al.
The Journal of pharmacology and experimental therapeutics, 228(3), 593-599 (1984-03-01)
Serum corticosterone concentrations were measured in rats after injection of fenfluramine (FEN), p-chloroamphetamine (PCA) and p-fluoroamphetamine (PFA), halogenated amphetamine derivatives believed to exert their behavioral and physiological effects through the release and/or depletion of brain serotonin (5-HT). Animals were pretreated
R E Dubin et al.
Progress in neuro-psychopharmacology & biological psychiatry, 9(5-6), 681-685 (1985-01-01)
(+)-Amphetamine (AM) and its fluorinated analogue (+)-2-amino-3-fluoro-1-phenylpropane (fluoroamphetamine, FAM) were compared with regard to their effects on locomotor and exploratory activity in mice. Both drugs caused a reduction in spontaneous exploration, but this effect was more marked with FAM than
D Marona-Lewicka et al.
European journal of pharmacology, 287(2), 105-113 (1995-12-12)
The present study was undertaken to compare the pharmacological properties of p-fluoroamphetamine with those of amphetamine and of other halogenated amphetamines, using several in vivo and in vitro tests. These included substitution testing in (+)-amphetamine (1 mg/kg, 5.4 mu mol/kg
T J Danielson et al.
Biochemical pharmacology, 35(24), 4423-4429 (1986-12-15)
The metabolism and some behavioral properties of each of the optical isomers of 2-amino-1-fluoro-3-phenylpropane hydrochloride (fluoroamphetamine, FAM) were examined and compared to those of the optical isomers of amphetamine (AM). Substitution of fluorine into the side-chain of AM increased the
Chih-Sheng Jhang et al.
Electrophoresis, 33(19-20), 3073-3078 (2012-09-25)
A novel drug-screening system, consisting of paper spray-MS (PS-MS) and a CE-ESI-MS method was developed. This system can be easily switched either to PS-MS for rapidly screening samples or to the traditional CE-ESI-MS method for separation and to obtain detailed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service