Skip to Content
Merck
All Photos(1)

Key Documents

E-052

Supelco

Ethylmorphine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23NO3
CAS Number:
Molecular Weight:
313.39
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CCOc1ccc2C[C@@H]3[C@@H]4C=C[C@H](O)[C@@H]5Oc1c2[C@]45CCN3C

InChI

1S/C19H23NO3/c1-3-22-15-7-4-11-10-13-12-5-6-14(21)18-19(12,8-9-20(13)2)16(11)17(15)23-18/h4-7,12-14,18,21H,3,8-10H2,1-2H3/t12-,13+,14-,18-,19-/m0/s1

InChI key

OGDVEMNWJVYAJL-LEPYJNQMSA-N

General description

Ethylmorphine is an opioid analgesic metabolized by the liver into morphine. The drug carries a risk of dependence and abuse. This certified solution standard is suited for LC/MS or GC/MS methods in forensic analysis, clinical toxicology, and urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gyula Szucs et al.
Chemico-biological interactions, 147(2), 151-161 (2004-03-12)
Tiamulin, a diterpene antibiotic, is used for treatment of pulmonary and gastrointestinal infections in swine and poultry. Combined administration of tiamulin and ionophores (e.g. monensin) to farm animals may lead to intoxication manifested in severe clinical symptoms. Tiamulin metabolite complex
K Hedenmalm et al.
Therapeutic drug monitoring, 19(6), 643-649 (1998-01-08)
The formation of morphine from codeine and ethylmorphine is mainly mediated by the polymorphic enzyme CYP2D6. The objective of this study was to investigate whether CYP2D6 poor metabolizers (PM) and CYP2D6 extensive metabolizers (EM) would respond differently during testing for
C Popa et al.
Journal of analytical toxicology, 22(2), 142-147 (1998-04-21)
In drugs-of-abuse testing, opiates constitute a delicate and controversial task because morphine may occur in urine for several reasons. Ethylmorphine (EtM), which is used as an antitussive drug in many countries, is metabolically converted to morphine. The present study was
A Wayne Jones et al.
Journal of analytical toxicology, 32(4), 265-272 (2008-04-24)
Morphine and codeine are frequently identified in blood samples from impaired drivers. But whether these opiates reflect the use of prescription analgesics or abuse of the illicit drug heroin (diacetyl morphine) is not always obvious. Opiates, either alone or together
S B Jia et al.
Hunan yi ke da xue xue bao = Hunan yike daxue xuebao = Bulletin of Hunan Medical University, 26(3), 215-216 (2003-01-23)
To find a reasonable method for treating and preventing fibrinous membrane after intraocular lens (IOL) implantation by using diclofenac sodium or dionine topically. Forty-eight eyes of twenty-four adult pure bred New Zealand rabbits were divided into three groups randomly. Group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service