Skip to Content
Merck
All Photos(1)

Key Documents

D-091

Supelco

10,11-Dihydro-10-Hydroxycarbamazepine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H14N2O2
CAS Number:
Molecular Weight:
254.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC1C2=C(C=CC=C2)N(C(N)=O)C3=C(C=CC=C3)C1

InChI

1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)

InChI key

BMPDWHIDQYTSHX-UHFFFAOYSA-N

General description

Also known as monohydroxy carbamazepine (MHC), 10,11-Dihydro-10-Hydroxycarbamazepine is a pharmacologically-active, primary metabolite of oxcarbazepine in urine. Oxcarbazepine, sold under the trade name Trileptal®, is an anticonvulsant drug used primarily for the treatment of epilepsy along with reduction of anxiety symptoms and treatment of mood disorders. Suitable uses for this certified solution standard include as a starting material for calibrators and controls in LC/MS or GC/MS oxcarbazepine testing applications for clinical toxicology, urine drug testing or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Trileptal is a registered trademark of Novartis Corporation

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2.0 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Julia Rossmann et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 1000, 84-94 (2015-07-27)
Matrix effects that occur during quantitative measurement by liquid chromatography mass spectrometry specifically when using electrospray ionization are a widely recognized phenomenon. Sample matrix compounds affect the ionization process of the target analytes, lead to a low signal response, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service