Skip to Content
Merck
All Photos(1)

Key Documents

D-065

Supelco

Dehydroepiandrosterone 3-sulfate (DHEAS) sodium salt solution

1.0 mg/mL in methanol (as free sulfate), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Dehydroepiandrosterone 3-sulfate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28NaO5S
CAS Number:
Molecular Weight:
391.48
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free sulfate)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C([C@@]1(C)CC[C@@]23[H])CC[C@]1([C@](CC=C4C[C@H](CC[C@]34C)OS([O-])(=O)=O)2[H])[H].[Na+]

InChI

1S/C19H28O5S.Na/c1-18-9-7-13(24-25(21,22)23)11-12(18)3-4-14-15-5-6-17(20)19(15,2)10-8-16(14)18;/h3,13-16H,4-11H2,1-2H3,(H,21,22,23);/q;+1/p-1/t13-,14-,15-,16-,18-,19-;/m0./s1

InChI key

GFJWACFSUSFUOG-ZJTJBYBXSA-M

General description

DHEAS is a metabolite of dehydroepiandrosterone (DHEA), an endogenous steroid that has been implicated in a broad range of biological effects in humans and other mammals. DHEAS levels are measured by LC-MS/MS for determination of excess adrenal activity as observed in adrenal cancer, certain forms of congenital adrenal hyperplasia (CAH), and polycystic ovary syndrome.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service