Skip to Content
Merck
All Photos(1)

Key Documents

C-109

Supelco

Catecholamine Mix 1 (Epinephrines) solution

1.0 mg/mL each component in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Epinephrines solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO3·HCl
Molecular Weight:
205.64
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®/SNAP-N-SHOOT®

composition

(±)-Epinephrine HCl
(±)-Norepinephrine HCl

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL each component in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing
forensics and toxicology

format

multi-component solution

storage temp.

−70°C

General description

A Certified Spiking Solution® of (±)-Epinephrine HCl and (±)-Norepinephrine HCl. This multi-component mix is suitable for use in preparation of linearity standards, calibrators, or controls in addition to other critical quantitative applications such as internal proficiency testing or OEM use in preparation of reagents, assay development, and instrument IQ/OQ. Certified concentration values for each component provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    (±)-Epinephrine hydrochloride
    DL-Norepinephrine hydrochloride

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service