Skip to Content
Merck
All Photos(1)

Key Documents

C-046

Supelco

Cannabinol solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26O2
CAS Number:
Molecular Weight:
310.43
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
cannabis testing

format

single component solution

storage temp.

−20°C

SMILES string

CCCCCc1cc(O)c-2c(OC(C)(C)c3ccc(C)cc-23)c1

InChI

1S/C21H26O2/c1-5-6-7-8-15-12-18(22)20-16-11-14(2)9-10-17(16)21(3,4)23-19(20)13-15/h9-13,22H,5-8H2,1-4H3

InChI key

VBGLYOIFKLUMQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The Certified Spiking Solution® is suitable for cannabinol testing by HPLC, GC/MS or LC-MS/MS methods in forensic analysis, clinical toxicology, urine drug testing, and Cannabis potency or impurity profiling testing applications.

Cannabinol is a phytocannabinoid that belongs to the class of C21 terpenophenolic compounds. Found in the cannabis plant it is a metabolite of Δ9-THC, the main psychoactive component of cannabis. Cannabinol is also the primary product of THC degradation from exposure to air and light.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:
  • Estimation of four cannabinoids from an oil matrix using reversed-phase high-performance liquid chromatography (RP-HPLC) combined with a diode array detector (DAD)
  • Simultaneous analysis of 11 cannabinoids in biomass and extracts of different cannabis varieties by an HPLC-UV method, following the International Conference on Harmonization (ICH) Tripartite Guideline for Validation of Analytical Procedures
  • Determination of eight cannabinoids in eight hemp samples and 12 forensic cannabis samples using reversed phase-high-pressure liquid chromatography (RP-HPLC) in combination with UV detector
  • Analysis of dried marijuana flowers to measure the amount of nine cannabinoids by high-performance liquid chromatography (HPLC) along with UV detection
  • Development and validation of an ultra high-performance liquid chromatographic (UHPLC) method coupled to photodiode array and mass spectrometry detectors to determine 11 cannabinoids in various Cannabis Sativa samples
  • Quantitative analysis of 17 cannabinoids in cannabis and hemp samples by LC-MS/MS following their liquid-solid extraction following AOAC and ASTM guidelines for cannabis and hemp matrices

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michelle L Holland et al.
European journal of pharmacology, 591(1-3), 128-131 (2008-07-16)
The ATP-binding cassette (ABC) transporter ABCC1, or multidrug resistance-related protein 1 (MRP1) is implicated in Phase II metabolism and multidrug resistance as it effluxes substrate anticancer drugs. As cannabinoids inhibit two related ABC transporters, P-glycoprotein and ABCG2, here we examined
Dayong Lee et al.
Clinical chemistry, 57(8), 1127-1136 (2011-06-17)
Oral fluid (OF) is an accepted alternative biological matrix for drug treatment, workplace, and DUID (driving under the influence of drugs) investigations, but establishing the cannabinoid OF detection window and concentration cutoff criteria are important. Cannabinoid concentrations were quantified in
Spyros P Nikas et al.
Journal of medicinal chemistry, 53(19), 6996-7010 (2010-10-12)
In pursuit of a more detailed understanding of the structural requirements for the key side chain cannabinoid pharmacophore, we have extended our SAR to cover a variety of conformationally modified side chains within the 9-keto and 9-hydroxyl tricyclic structures. Of
Satoshi Yamaori et al.
Biochemical pharmacology, 79(11), 1691-1698 (2010-02-02)
Inhibitory effects of Delta(9)-tetrahydrocannabinol (Delta(9)-THC), cannabidiol (CBD), and cannabinol (CBN), the three major constituents in marijuana, on catalytic activities of human cytochrome P450 (CYP) 1 enzymes were investigated. These cannabinoids inhibited 7-ethoxyresorufin O-deethylase activity of recombinant CYP1A1, CYP1A2, and CYP1B1
F Van der Kooy et al.
Inhalation toxicology, 21(2), 87-90 (2008-10-16)
Medicinal cannabis has attracted a lot of attention in recent times. Various forms of administration are used, of which smoking is very common but the least desirable. Smoking cannabis generates a large amount of unwanted side products, of which carcinogenic

Articles

ISO-certified cannabinoid CRMs, THC and THCA, available as single components or mixes, for accurate potency testing.

ISO-certified cannabinoid CRMs, THC and THCA, available as single components or mixes, for accurate potency testing.

ISO-certified cannabinoid CRMs, THC and THCA, available as single components or mixes, for accurate potency testing.

ISO-certified cannabinoid CRMs, THC and THCA, available as single components or mixes, for accurate potency testing.

See All

Protocols

Rapid potency testing of marijuana-infused edibles using LC/MS on a biphenyl stationary phase detected eleven cannabinoids.

Rapid potency testing of marijuana-infused edibles using LC/MS on a biphenyl stationary phase detected eleven cannabinoids.

Rapid potency testing of marijuana-infused edibles using LC/MS on a biphenyl stationary phase detected eleven cannabinoids.

Rapid potency testing of marijuana-infused edibles using LC/MS on a biphenyl stationary phase detected eleven cannabinoids.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service