Skip to Content
Merck
All Photos(1)

Key Documents

A-923

Supelco

Amitriptyline hydrochloride solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24ClN
CAS Number:
Molecular Weight:
313.86
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CN(C)CC\C=C1\c2ccccc2CCc3ccccc13

InChI

1S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12H,7,13-15H2,1-2H3;1H

InChI key

KFYRPLNVJVHZGT-UHFFFAOYSA-N

Gene Information

General description

A Certified Spiking Solution® suitable for use in LC/MS or GC/MS applications in clinical toxicology, forensic analysis, urine drug testing, prescription monitoring or pharmaceutical research. Amitriptyline, marketed as the brand drug Elavil, Tryptizol, or Laroxyl, is a tricyclic antidepressant used in the treatment of major depression, clinical/endogenous depression and involutional melancholia.

Application


  • Tricyclic antidepressant research: A study demonstrated the use of antisolvent precipitation to formulate excipient-free nanoparticles of psychotropic drugs, including amitriptyline hydrochloride, which could significantly impact the development of novel drug delivery systems for neuropharmacological applications (Wu and Wang, 2022).

  • Neuropharmacology amitriptyline: Research into micellar and interfacial phenomena of amitriptyline hydrochloride with cationic ester-bonded gemini surfactants explored its potential in enhancing drug solubility and stability, which is crucial for neuropharmacological studies and drug delivery systems (Abdul Rub, 2020).

  • Amitriptyline liquid formulation: A methodology to create polymeric nanocarriers containing hydrophilic low molecular-weight drugs, such as amitriptyline, offers a green strategy with very high drug loading, facilitating the development of amitriptyline-based formulations for pharmacological research (Villamizar-Sarmiento et al., 2019).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Sweating under polypharmacy with tramadol, amitriptyline and duloxetine?].
Gesine Picksak et al.
Medizinische Monatsschrift fur Pharmazeuten, 36(5), 195-196 (2013-06-14)
David Temesi et al.
Journal of pharmaceutical and biomedical analysis, 76, 164-168 (2013-01-22)
Procedures for drug monitoring based on Dried Blood Spot (DBS) sampling are gaining acceptance for an increasing number of clinical and preclinical applications, where ease of use, small sample requirement, and improved sample stability have been shown to offer advantages
Topical amitriptyline-ketamine for the treatment of brachioradial pruritus.
Timothy J Poterucha et al.
JAMA dermatology, 149(2), 148-150 (2013-02-22)
Christian Janfelt et al.
Journal of mass spectrometry : JMS, 48(3), 361-366 (2013-03-16)
Displaced dual-mode imaging (DDI) is introduced as a method for simultaneous imaging in positive and negative-ion mode on the same sample with desorption electrospray ionization imaging, as well as a method for simultaneous imaging in full-scan and tandem mass spectrometry
Recreational amitriptyline toxicity mimicking basilar artery stroke.
Jonathan Birns et al.
European journal of emergency medicine : official journal of the European Society for Emergency Medicine, 20(2), 139-140 (2013-02-26)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service