Skip to Content
Merck
All Photos(1)

Key Documents

LM1500

Avanti

12:0-13:0 PI

Avanti Research - A Croda Brand LM1500, methanol solution

Synonym(s):

1-dodecanoyl-2-tridecanoyl-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H68NO13P
CAS Number:
Molecular Weight:
729.88
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1500-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1500

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(OC(CCCCCCCCCCCC)=O)COC(CCCCCCCCCCC)=O.[NH4+]

InChI key

REGULBPHXGUPTE-RBKCQXOZSA-N

General description

Phosphatidylinositol (PI) is commonly found in animal tissues and bacteria. PI is soluble in water and chloroform/methanol.[1]

Application

12:0-13:0 PI or 1-dodecanoyl-2-tridecanoyl-sn-glycero-3-phospho-(1′-myo-inositol) has been used as an internal standard for lipid quantification/analysis using electrospray ionization tandem mass spectrometry (ESI-MS/MS)[2] and liquid chromatography mass spectrometry (LC-MS).[3][4][5]

Packaging

2 mL Amber Glass Sealed Ampule (LM1500-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Penelope Dimas et al.
eLife, 8 (2019-05-08)
Oligodendrocytes (OLs) support neurons and signal transmission in the central nervous system (CNS) by enwrapping axons with myelin, a lipid-rich membrane structure. We addressed the significance of fatty acid (FA) synthesis in OLs by depleting FA synthase (FASN) from OL
Laura Montani et al.
The Journal of cell biology, 217(4), 1353-1368 (2018-02-13)
Myelination calls for a remarkable surge in cell metabolism to facilitate lipid and membrane production. Endogenous fatty acid (FA) synthesis represents a potentially critical process in myelinating glia. Using genetically modified mice, we show that Schwann cell (SC) intrinsic activity
Ulrike Bruning et al.
Cell metabolism, 28(6), 866-880 (2018-08-28)
The role of fatty acid synthesis in endothelial cells (ECs) remains incompletely characterized. We report that fatty acid synthase knockdown (FASNKD) in ECs impedes vessel sprouting by reducing proliferation. Endothelial loss of FASN impaired angiogenesis in vivo, while FASN blockade reduced
Milena Vukotic et al.
Molecular cell, 67(3), 471-483 (2017-07-18)
Mutations in mitochondrial acylglycerol kinase (AGK) cause Sengers syndrome, which is characterized by cataracts, hypertrophic cardiomyopathy, and skeletal myopathy. AGK generates phosphatidic acid and lysophosphatidic acid, bioactive phospholipids involved in lipid signaling and the regulation of tumor progression. However, the
Mouhannad Malek et al.
Molecular cell, 68(3), 566-580 (2017-10-24)
The PI3K signaling pathway regulates cell growth and movement and is heavily mutated in cancer. Class I PI3Ks synthesize the lipid messenger PI(3,4,5)P3. PI(3,4,5)P3 can be dephosphorylated by 3- or 5-phosphatases, the latter producing PI(3,4)P2. The PTEN tumor suppressor is thought

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service