Skip to Content
Merck
All Photos(1)

Key Documents

LM1004

Avanti

17:0-14:1 PC

Avanti Research - A Croda Brand LM1004, methanol solution

Synonym(s):

1-heptadecanoyl-2-(9Z-tetradecenoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H76NO8P
CAS Number:
Molecular Weight:
718.00
UNSPSC Code:
12352211
NACRES:
NA.25

form

methanol solution

packaging

pkg of 1 × 1 mL (LM1004-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM1004

concentration

~10 μg/mL (Refer to C of A for lot specific concentration.)

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCC)=O)COC(CCCCCCCCCCCCCCCC)=O)=O

Application

17:0-14:1 PC or 1-heptadecanoyl-2-(9Z-tetradecenoyl)-sn-glycero-3-phosphocholine has been used as an internal standard:
  • for phospholipid dipalmitoylphosphatidylcholine (DPPC), quantification by selected reaction monitoring method
  • for lipid analysis of murine brain lipid extract using Fourier transform-ion cyclotron resonance-mass spectrometry (FT-ICR-MS)
  • in lipid extraction to control extraction efficiency, evaluate liquid chromatography (LC)-MS performance and normalize the LC-MS data
  • for lipid quantification using LC-MS

Packaging

2 mL Amber Glass Sealed Ampule (LM1004-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kévin Contrepois et al.
Scientific reports, 8(1), 17747-17747 (2018-12-12)
Lipidomics - the global assessment of lipids - can be performed using a variety of mass spectrometry (MS)-based approaches. However, choosing the optimal approach in terms of lipid coverage, robustness and throughput can be a challenging task. Here, we compare
Andreas Ullen et al.
Free radical biology & medicine, 49(11), 1655-1665 (2010-09-03)
Plasmalogens, 1-O-alk-1'-enyl-2-acyl-sn-glycerophospholipids, are significant constituents of cellular membranes and are essential for normal brain development. Plasmalogens, which contain a vinyl ether bond at the sn-1 position, are preferential targets for hypochlorous acid (HOCl), generated by myeloperoxidase (MPO) from H(2)O(2) and
Per Larsson et al.
Respiratory physiology & neurobiology, 243, 39-46 (2017-05-16)
Exhaled particles constitute a micro-sample of respiratory tract lining fluid. Inhalations from low lung volumes generate particles in small airways by the airway re-opening mechanism. Forced exhalations are assumed to generate particles in central airways by mechanisms associated with high
Milena Vukotic et al.
Molecular cell, 67(3), 471-483 (2017-07-18)
Mutations in mitochondrial acylglycerol kinase (AGK) cause Sengers syndrome, which is characterized by cataracts, hypertrophic cardiomyopathy, and skeletal myopathy. AGK generates phosphatidic acid and lysophosphatidic acid, bioactive phospholipids involved in lipid signaling and the regulation of tumor progression. However, the
Alexander Fauland et al.
Journal of lipid research, 52(12), 2314-2322 (2011-10-01)
This work aims to combine chromatographic retention, high mass resolution and accuracy, MS/MS spectra, and a package for automated identification and quantitation of lipid species in one platform for lipidomic analysis. The instrumental setup elaborated comprises reversed-phase HPLC coupled to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service