Skip to Content
Merck
All Photos(1)

Key Documents

880123P

Avanti

DSPE-PEG(5000) Folate

Avanti Research - A Croda Brand 880123P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-5000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 500 μg (880123P-500ug)

manufacturer/tradename

Avanti Research - A Croda Brand 880123P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC([(OCH2CH2)113]NC(CC[C@H](NC(C1=CC=C(NCC(C=N2)=NC3=C2N=C(N)NC3=O)C=C1)=O)C([O-])=O)=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+].[NH4+]

General description

DSPE-PEG(5000) Folate is a conjugate of folic acid and amino-PEG-DSPE (1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-(polyethylene glycol).

Biochem/physiol Actions

Folate receptors are cellular surface markers for numerous solid tumors. DSPE-PEG-folate (1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[folate(polyethylene glycol)-5000]) incorporated into liposomes serves as a targeting ligand. Folate targeted drug delivery is useful in diagnosing and treating cancer and inflammatory diseases. Many molecules such as toxins (proteinaceous), immune stimulators and anti-cancer drugs can be delivered to folate receptor.

Packaging

5 mL Amber Glass Screw Cap Vial (880123P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bioinspired and Biomimetic Polymer Systems for Drug and Gene Delivery, 48-48 (2014)
Folate receptor-mediated drug targeting: from therapeutics to diagnostics
Hilgenbrink AR and Low PS
Journal of Pharmaceutical Sciences, 94(10), 2135-2146 (2005)
Folate-targeted gadolinium-lipid-based nanoparticles as a bimodal contrast agent for tumor fluorescent and magnetic resonance imaging
Nakamura T, et al.
Biological & Pharmaceutical Bulletin, 37(4), 521-527 (2014)
Shih-Jiuan Chiu et al.
Journal of controlled release : official journal of the Controlled Release Society, 112(2), 199-207 (2006-03-28)
A novel transferrin receptor (TfR)-targeted liposomal formulation was synthesized and evaluated for the delivery of a phosphorothioate antisense oligodeoxyribonucleotide (ODN) (G3139, oblimerson sodium, or Genasense) to Bcl-2 in K562 leukemia cells. Liposomes composed of DC-Chol/egg PC/PEG-DSPE (25:73.5:1.5, mol/mol/mol) were loaded
A Gabizon et al.
Bioconjugate chemistry, 10(2), 289-298 (1999-03-17)
Conjugates of three components, folic acid-poly(ethylene glycol)-distearoylphosphatidylethanolamine (FA-PEG-DSPE), derived from PEG with molecular masses of 2000 and 3350 Da were synthesized by a carbodiimide-mediated coupling of FA to H2N-PEG-DSPE. The conjugates were characterized by 1H NMR, MALDI-TOF, and HPLC analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service