Skip to Content
Merck
All Photos(1)

Key Documents

870608H

Avanti

4-HNE-dimethylacetal

Avanti Research - A Croda Brand 870608H, hexane solution

Synonym(s):

(E)-4R-hydroxynonenal-dimethylacetal (Each vial contains 6.8mg which yields ~5.2mg 4-HNE after deprotection)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22O3
CAS Number:
Molecular Weight:
202.29
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

hexane solution

packaging

pkg of 1 × 1 mL (870608H-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870608H

concentration

6.8 mg/mL (870608H-5mg)

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCC[C@@H](O)/C=C/C(OC)OC

InChI key

ATGIHMSJAARJTF-AAXQSMANSA-N

General description

4-Hydroxynonenal (4-HNE or HNE) is an α,β-unsaturated hydroxyalkenal which is produced by lipid peroxidation in cells. 4-HNE has been hypothesized by several researchers to play a key role in cell signal transduction, in a variety of pathways from cell cycle events to cellular adhesion.
The dimethylacetal derivative is a more stable form of 4-HNE which is good for long term storage of the compound during use. It can be easily converted to 4-HNE in the presence of acid.

Packaging

2 mL Amber Glass Sealed Ampule (870608H-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

On the role of 4-hydroxynonenal in health and disease
Csala M, et al.
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease, 1852(5), 826-838 (2015)
Kamelija Zarkovic
Molecular aspects of medicine, 24(4-5), 293-303 (2003-08-02)
The development of oxidative stress, in which production of highly reactive oxygen species (ROS) overwhelms antioxidant defenses, is a feature of many neurological diseases: ischemic, inflammatory, metabolic and degenerative. Oxidative stress is increasingly implicated in a number of neurodegenerative disorders
Neven Zarkovic
Molecular aspects of medicine, 24(4-5), 281-291 (2003-08-02)
The review is focused on the currently major aspect of 4-hydroxynonenal (HNE) research--studies that combine biological activities of the aldehyde together with the methods of its identification in cells and tissues. Because there were some excellent reviews on HNE published
Yogesh C Awasthi et al.
Free radical biology & medicine, 37(5), 607-619 (2004-08-04)
4-Hydroxynonenal (HNE), one of the major end products of lipid peroxidation, has been shown to be involved in signal transduction and available evidence suggests that it can affect cell cycle events in a concentration-dependent manner. Glutathione S-transferases (GSTs) can modulate
N Zarković et al.
Life sciences, 65(18-19), 1901-1904 (1999-11-27)
Immunohistochemical analysis of the distribution of the lipid peroxidation product 4-hydroxynonenal (HNE) in the brain of baboons exposed to experimental hemorrhagic traumatic shock or sepsis showed that systemic oxidative stress and the thereby generated HNE affect the blood:brain barrier and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service