Skip to Content
Merck
All Photos(1)

Key Documents

860699P

Avanti

C17:0 GB3 (synthetic)

powder

Synonym(s):

C17:0 Globotriaosylceramide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H99NO18
CAS Number:
Molecular Weight:
1038.35
UNSPSC Code:
12352211
NACRES:
NA.28

Assay

99% (TLC)

form

powder

packaging

package of 1 × 500 μg (860699P-500UG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](OC[C@H](NC(CCCCCCCCCCCCCCCC)=O)[C@H](O)/C=C/CCCCCCCCCCCCC)O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O)[C@H]2O)O[C@H]3O[C@H](CO)[C@@H]([C@H](O)[C@H]3O)O)[C@@H]1O

General description

Globotriaosylceramide (GB3) is a glycosphingolipid. It is located in the cell membrane of endothelial cell of glomeruli and human erythrocytes.

Biochem/physiol Actions

Globotriaosylceramide (GB3) is a potent receptor for verotoxin. It is expressed at high concentrations in Burkitt lymphoma cell lines, human embryonal carcinoma and other tumor cells.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (860699P-500UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of globotriaosylceramide (Gb3) and lysoglobotriaosylceramide (lysoGb3)
Nicolaou K C, et al.
Carbohydrate Research, 202, 177-191 (1990)
Modelling of the interaction of verotoxin-1 (VT1) with its glycolipid receptor, globotriaosylceramide (Gb3)
Nyholm P G, et al.
International Journal of Biological Macromolecules, 17(3-4), 199-204 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service