Skip to Content
Merck
All Photos(1)

Key Documents

860495P

Avanti

Monomethyl Sphingosine (d18:1)

Avanti Research - A Croda Brand 860495P, powder

Synonym(s):

N-methyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H39NO2
CAS Number:
Molecular Weight:
313.52
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860495P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860495P

lipid type

deuterated lipids
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](NC)([H])[C@]([H])(O)/C=C/CCCCCCCCCCCCC

General description

Monomethyl sphingosine or N-methyl-D-erythro-sphingosine is a N-methyl derivative of sphingosine

Application

Monomethyl sphingosine or N-methyl-D-erythro-sphingosine may be used as one of the substrates for sphingosine methyltransferase to test its substrate specificity in rat kidney homogenate.

Biochem/physiol Actions

Monomethyl sphingosine inhibits protein kinase C. It also modulates other protein kinases and enzymes involved in signal transduction.

Packaging

5 mL Amber Glass Screw Cap Vial (860495P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biological Function of Gangliosides (1994)
Handbook of Neurochemistry and Molecular Neurobiology:Neural Lipids (2009)
Santosh J Sacket et al.
Acta pharmacologica Sinica, 29(10), 1227-1232 (2008-09-27)
To characterize sphingosine methyltransferase in rat tissues. By using S-adenosyl-L-(methyl-3H) methionine, enzymatic activity was measured in the rat liver and kidney homogenates. The optimum pH and reaction time for the enzyme assay were pH 7.8 and 1 h. ZnCl2 inhibited

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service