Skip to Content
Merck
All Photos(2)

Key Documents

858143P

Avanti

18:1 Lyso PS

Avanti Research - A Croda Brand

Synonym(s):

1-(9Z-octadecenoyl)-sn-glycero-3-phospho-L-serine (sodium salt); PS(18:1(9Z)/0:0); 18:1 LPS; C18:1 LPS; 110726

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H45NO9PNa
CAS Number:
Molecular Weight:
545.58
UNSPSC Code:
51191904
NACRES:
NA.25

description

1-oleoyl-2-hydroxy-sn-glycero-3-phospho-L-serine (sodium salt)

Assay

99% (LPS; may contain up to 15% of the 2-LPS isomer, TLC)

form

powder

packaging

pkg of 1 × 10 mg (858143P-10mg)
pkg of 1 × 100 mg (858143P-100mg)
pkg of 1 × 25 mg (858143P-25mg)
pkg of 1 × 5 mg (858143P-5mg)
pkg of 1 × 50 mg (858143P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OC[C@](C([O-])=O)([H])[NH3+])=O)([H])COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

Application

18:1 Lyso PS has been used:
  • for the preparation of phospholipid vesicles
  • to study its effects on nerve growth factor (NGF)-induced mitogen-activated protein kinase (MAPK) and serine/threonine-protein kinase (Akt) signals
  • to prevent the lack of de novo synthesis of phosphatidylserine (PS) in Golgi apparatus

Packaging

5 mL Clear Glass Sealed Ampule (858143P-100mg)
5 mL Clear Glass Sealed Ampule (858143P-10mg)
5 mL Clear Glass Sealed Ampule (858143P-25mg)
5 mL Clear Glass Sealed Ampule (858143P-50mg)
5 mL Clear Glass Sealed Ampule (858143P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The catalytic domains of Clostridium sordellii lethal toxin and related large clostridial glucosylating toxins specifically recognize the negatively charged phospholipids phosphatidylserine and phosphatidic acid
Varela Chavez C, et al.
Cellular Microbiology, 17(10), 1477-1493 (2015)
Suriakarthiga Ganesan et al.
Traffic (Copenhagen, Denmark), 20(3), 226-245 (2018-12-21)
Diacylglycerol (DAG) is a key signaling lipid and intermediate in lipid metabolism. Our knowledge of DAG distribution and dynamics in cell membranes is limited. Using live-cell fluorescence microscopy we investigated the localization of yeast cytosolic-facing pools of DAG in response
Takafumi Matsumura et al.
Scientific reports, 11(1), 3458-3458 (2021-02-12)
In vitro spermatogenesis (IVS) using air-liquid interphase organ culture method is possible with mouse testis tissues. The same method, however, has been hardly applicable to animals other than mice, only producing no or limited progression of spermatogenesis. In the present
Qingxin Shi et al.
Food & function, 10(12), 7782-7792 (2019-11-30)
Triterpenoid saponins from Kuding tea have demonstrated preventive effects on hyperlipidaemia induced by a high-fat diet. Lysoglycerophospholipids (Lyso-GPLs) are known to be associated with proatherogenic conditions such as hyperlipidaemia. In this study, a target profiling strategy based on a multiple
Jung Yong Eum et al.
Journal of chromatography. A, 460849-460849 (2020-01-14)
Aging refers to the intracellular accumulation of reactive oxygen species that damages proteins, DNA, and lipids. As alterations in lipid metabolism may trigger metabolic disorders and the onset of metabolic diseases, changes in lipid profiles can be closely related to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service