Skip to Content
Merck
All Photos(2)

Key Documents

857460M

Avanti

16:0 aldehyde-d9

hexadecanal(d9), methylene chloride solution

Synonym(s):

110843

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23OD9
CAS Number:
Molecular Weight:
249.48
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>90% (contains dimer, trimer, tetramer, TLC)

form

methylene chloride solution

packaging

pkg of 1 × 1 mL (857460M-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 857460M

concentration

1 mg/mL (857460M-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])(CCCCCCCCCCCC([H])=O)[2H]

General description

16:0 aldehyde-d9 or hexadecanal-d9 is a deuterated C16 chain length fatty aldehyde. Hexadecanal may be derived from a plasmalogen. Hexadecanal is reduced to hexadecanol by the reaction catalyzed by alcohol dehydrogenase, propanol-preferring (Adhp). Hexadecanol has wide applications in chemical & fuel industries.

Application

16:0 aldehyde-d9 has been used as a fatty aldehyde internal standard for derivatization and mass spectrometric identification of α-hydroxy fatty aldehydes.

Packaging

5 mL Clear Glass Sealed Ampule (857460M-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and quantitation of hexadecanal and octadecanal in broiler muscle phospholipids
Gardner HK, et al.
Poultry Science, 51(3), 1056-1058 (1972)
Identification of long chain specific aldehyde reductase and its use in enhanced fatty alcohol production in E. coli
Fatma Z, et al.
Metabolic engineering, 37, 35-45 (2016)
Cytochrome c is an oxidative stress-activated plasmalogenase that cleaves plasmenylcholine and plasmenylethanolamine at the sn-1 vinyl ether linkage
Jenkins CM, et al.
The Journal of Biological Chemistry, 293(22), 8693-8709 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service